Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 85846236-85846479 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ST3GAL5
Entrez Gene ID
8869
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 5
Gene Aliases
SATI, SIAT9, SIATGM3S, ST3GalV
UniGene
Hs.415117
Gene Chromosome Location
Chr.2: 85839148-85889034 on Build GRCh38
Transcripts
NM_001042437.1 NM_003896.3 XM_005264630.4 XM_011533143.2 XM_017005199.1 XM_017005200.1 XM_017005201.1 XM_017005202.1 XM_017005203.1 XM_017005204.1 XM_017005205.1 XM_017005206.1 XM_017005207.1 XM_017005208.1 XM_017005209.1 XM_017005210.1 XM_017005211.1 XM_017005212.1 XM_017005213.1 XM_017005214.1 XM_017005215.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6458763 c.770_771insA p.E258fs*40 Chr.2: 85846386-85846387 on Build GRCh38
6791088 c.744C>A p.F248L Chr.2: 85846413-85846413 on Build GRCh38
6791087 c.813C>A p.F271L Chr.2: 85846413-85846413 on Build GRCh38
6458762 c.839_840insA p.E281fs*40 Chr.2: 85846386-85846387 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751569436 Chr.2: 85846395-85846395 on Build GRCh38
rs994246159 Chr.2: 85846319-85846322 on Build GRCh38
rs766279321 Chr.2: 85846445-85846445 on Build GRCh38
rs544272877 Chr.2: 85846287-85846287 on Build GRCh38
rs114807360 Chr.2: 85846288-85846288 on Build GRCh38
rs372131685 Chr.2: 85846267-85846267 on Build GRCh38
rs772981914 Chr.2: 85846423-85846423 on Build GRCh38
rs150114705 Chr.2: 85846422-85846422 on Build GRCh38
rs768216151 Chr.2: 85846362-85846362 on Build GRCh38
rs1038571940 Chr.2: 85846389-85846389 on Build GRCh38
rs773953439 Chr.2: 85846363-85846363 on Build GRCh38
rs369127120 Chr.2: 85846426-85846426 on Build GRCh38
rs562660240 Chr.2: 85846331-85846331 on Build GRCh38
rs933992277 Chr.2: 85846284-85846284 on Build GRCh38
rs748775245 Chr.2: 85846360-85846360 on Build GRCh38
rs761427979 Chr.2: 85846411-85846411 on Build GRCh38
rs752738939 Chr.2: 85846448-85846448 on Build GRCh38
rs80130823 Chr.2: 85846358-85846358 on Build GRCh38
rs780237380 Chr.2: 85846342-85846342 on Build GRCh38
rs889008859 Chr.2: 85846272-85846272 on Build GRCh38
rs146939269 Chr.2: 85846449-85846449 on Build GRCh38
rs753760726 Chr.2: 85846281-85846281 on Build GRCh38
rs199703454 Chr.2: 85846402-85846402 on Build GRCh38

View Full Product Details