Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 48979433-48979688 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WNT1
Entrez Gene ID
7471
Gene Name
Wnt family member 1
Gene Aliases
BMND16, INT1, OI15
UniGene
Hs.248164
Gene Chromosome Location
Chr.12: 48978453-48982613 on Build GRCh38
Transcripts
NM_005430.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5155255 c.295A>G p.N99D Chr.12: 48979658-48979658 on Build GRCh38
4042309 c.319G>A p.A107T Chr.12: 48979682-48979682 on Build GRCh38
5841093 c.149delC p.K51fs*14 Chr.12: 48979512-48979512 on Build GRCh38
4042308 c.302G>T p.R101L Chr.12: 48979665-48979665 on Build GRCh38
6072537 c.125C>T p.S42F Chr.12: 48979488-48979488 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778294620 Chr.12: 48979612-48979613 on Build GRCh38
rs775138924 Chr.12: 48979541-48979541 on Build GRCh38
rs774290977 Chr.12: 48979597-48979597 on Build GRCh38
rs753162212 Chr.12: 48979512-48979512 on Build GRCh38
rs780679366 Chr.12: 48979651-48979651 on Build GRCh38
rs766584422 Chr.12: 48979474-48979474 on Build GRCh38
rs770381881 Chr.12: 48979534-48979534 on Build GRCh38
rs977301761 Chr.12: 48979578-48979578 on Build GRCh38
rs61758378 Chr.12: 48979627-48979627 on Build GRCh38
rs779414153 Chr.12: 48979509-48979509 on Build GRCh38
rs764154512 Chr.12: 48979458-48979458 on Build GRCh38
rs758809246 Chr.12: 48979517-48979517 on Build GRCh38
rs1033810883 Chr.12: 48979625-48979625 on Build GRCh38
rs755253307 Chr.12: 48979482-48979482 on Build GRCh38
rs765605638 Chr.12: 48979618-48979618 on Build GRCh38
rs750451481 Chr.12: 48979461-48979461 on Build GRCh38
rs757767948 Chr.12: 48979634-48979634 on Build GRCh38
rs754052778 Chr.12: 48979481-48979481 on Build GRCh38
rs769504937 Chr.12: 48979537-48979537 on Build GRCh38
rs1027727196 Chr.12: 48979515-48979515 on Build GRCh38
rs200097796 Chr.12: 48979613-48979613 on Build GRCh38
rs375172611 Chr.12: 48979560-48979560 on Build GRCh38
rs887278549 Chr.12: 48979642-48979642 on Build GRCh38
rs762859463 Chr.12: 48979544-48979544 on Build GRCh38
rs993620313 Chr.12: 48979478-48979478 on Build GRCh38
rs1048999636 Chr.12: 48979629-48979629 on Build GRCh38
rs761666594 Chr.12: 48979598-48979598 on Build GRCh38
rs958164260 Chr.12: 48979480-48979480 on Build GRCh38
rs144907447 Chr.12: 48979535-48979535 on Build GRCh38
rs769418812 Chr.12: 48979662-48979662 on Build GRCh38
rs768363275 Chr.12: 48979590-48979590 on Build GRCh38
rs912508879 Chr.12: 48979585-48979585 on Build GRCh38
rs181106181 Chr.12: 48979522-48979522 on Build GRCh38
rs766496538 Chr.12: 48979612-48979612 on Build GRCh38
rs747502483 Chr.12: 48979523-48979523 on Build GRCh38
rs760763815 Chr.12: 48979462-48979462 on Build GRCh38
rs1039388659 Chr.12: 48979609-48979609 on Build GRCh38
rs745442968 Chr.12: 48979654-48979654 on Build GRCh38
rs369711501 Chr.12: 48979630-48979630 on Build GRCh38

View Full Product Details