Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 149496726-149496993 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSNK1A1
Entrez Gene ID
1452
Gene Name
casein kinase 1 alpha 1
Gene Aliases
CK1, CK1a, CKIa, HEL-S-77p, HLCDGP1, PRO2975
UniGene
Hs.529862
Gene Chromosome Location
Chr.5: 149495894-149551552 on Build GRCh38
Transcripts
NM_001025105.2 NM_001271741.1 NM_001271742.1 NM_001892.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5356362 c.1091-6delT p.? Chr.5: 149496866-149496866 on Build GRCh38
5356363 c.1007-6delT p.? Chr.5: 149496866-149496866 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756936113 Chr.5: 149496879-149496879 on Build GRCh38
rs745857376 Chr.5: 149496887-149496888 on Build GRCh38
rs751956369 Chr.5: 149496846-149496846 on Build GRCh38
rs750104211 Chr.5: 149496817-149496817 on Build GRCh38
rs936279713 Chr.5: 149496890-149496890 on Build GRCh38
rs777162478 Chr.5: 149496806-149496808 on Build GRCh38
rs575499750 Chr.5: 149496921-149496921 on Build GRCh38
rs769926155 Chr.5: 149496882-149496882 on Build GRCh38
rs914796549 Chr.5: 149496934-149496934 on Build GRCh38
rs201469252 Chr.5: 149496880-149496880 on Build GRCh38
rs749520802 Chr.5: 149496894-149496894 on Build GRCh38
rs777893488 Chr.5: 149496886-149496886 on Build GRCh38
rs1030145443 Chr.5: 149496872-149496872 on Build GRCh38
rs780936669 Chr.5: 149496878-149496879 on Build GRCh38
rs188932819 Chr.5: 149496796-149496796 on Build GRCh38
rs941398630 Chr.5: 149496795-149496795 on Build GRCh38
rs912284175 Chr.5: 149496930-149496930 on Build GRCh38
rs746624658 Chr.5: 149496876-149496878 on Build GRCh38
rs543086891 Chr.5: 149496961-149496961 on Build GRCh38
rs758049682 Chr.5: 149496826-149496826 on Build GRCh38
rs376180123 Chr.5: 149496966-149496966 on Build GRCh38
rs781670961 Chr.5: 149496863-149496863 on Build GRCh38
rs780326709 Chr.5: 149496840-149496840 on Build GRCh38
rs755283157 Chr.5: 149496850-149496850 on Build GRCh38
rs761414239 Chr.5: 149496814-149496814 on Build GRCh38
rs942438490 Chr.5: 149496932-149496932 on Build GRCh38
rs974953912 Chr.5: 149496851-149496851 on Build GRCh38
rs913691889 Chr.5: 149496873-149496873 on Build GRCh38
rs764800978 Chr.5: 149496815-149496815 on Build GRCh38
rs771153653 Chr.5: 149496898-149496898 on Build GRCh38
rs916568538 Chr.5: 149496773-149496773 on Build GRCh38

View Full Product Details