Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7465607-7465864 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZBTB4
Entrez Gene ID
57659
Gene Name
zinc finger and BTB domain containing 4
Gene Aliases
KAISO-L1, ZNF903
UniGene
Hs.35096
Gene Chromosome Location
Chr.17: 7459366-7484249 on Build GRCh38
Transcripts
NM_001128833.1 NM_020899.3 XM_006721563.3 XM_006721564.2 XM_011523972.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
385429 c.1027G>C p.E343Q Chr.17: 7465775-7465775 on Build GRCh38
4139824 c.1065T>C p.H355H Chr.17: 7465737-7465737 on Build GRCh38
1289009 c.1010A>C p.Y337S Chr.17: 7465792-7465792 on Build GRCh38
3932733 c.1009T>G p.Y337D Chr.17: 7465793-7465793 on Build GRCh38
3680559 c.1032A>G p.K344K Chr.17: 7465770-7465770 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750537520 Chr.17: 7465663-7465663 on Build GRCh38
rs756609356 Chr.17: 7465699-7465699 on Build GRCh38
rs377098416 Chr.17: 7465743-7465743 on Build GRCh38
rs775155255 Chr.17: 7465755-7465755 on Build GRCh38
rs551738890 Chr.17: 7465645-7465645 on Build GRCh38
rs999930250 Chr.17: 7465657-7465657 on Build GRCh38
rs201264238 Chr.17: 7465792-7465792 on Build GRCh38
rs777368119 Chr.17: 7465708-7465708 on Build GRCh38
rs560386263 Chr.17: 7465714-7465714 on Build GRCh38
rs753390026 Chr.17: 7465847-7465847 on Build GRCh38
rs900708755 Chr.17: 7465806-7465806 on Build GRCh38
rs918493239 Chr.17: 7465695-7465695 on Build GRCh38
rs891643772 Chr.17: 7465664-7465664 on Build GRCh38
rs533527749 Chr.17: 7465690-7465690 on Build GRCh38
rs763061413 Chr.17: 7465794-7465794 on Build GRCh38
rs763117108 Chr.17: 7465674-7465674 on Build GRCh38
rs781539599 Chr.17: 7465692-7465692 on Build GRCh38
rs751868936 Chr.17: 7465800-7465800 on Build GRCh38
rs139137598 Chr.17: 7465716-7465716 on Build GRCh38
rs766638056 Chr.17: 7465683-7465683 on Build GRCh38
rs767882248 Chr.17: 7465802-7465802 on Build GRCh38
rs143028324 Chr.17: 7465830-7465830 on Build GRCh38
rs778772638 Chr.17: 7465722-7465722 on Build GRCh38
rs753178934 Chr.17: 7465693-7465693 on Build GRCh38
rs1018871555 Chr.17: 7465698-7465698 on Build GRCh38
rs771844823 Chr.17: 7465748-7465748 on Build GRCh38
rs151017355 Chr.17: 7465770-7465770 on Build GRCh38
rs569544436 Chr.17: 7465783-7465783 on Build GRCh38
rs756667255 Chr.17: 7465815-7465815 on Build GRCh38
rs200433979 Chr.17: 7465691-7465691 on Build GRCh38
rs890406881 Chr.17: 7465686-7465686 on Build GRCh38
rs765237689 Chr.17: 7465662-7465662 on Build GRCh38
rs759680223 Chr.17: 7465801-7465801 on Build GRCh38

View Full Product Details