Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 131085465-131085722 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP36
Entrez Gene ID
158763
Gene Name
Rho GTPase activating protein 36
Gene Aliases
-
UniGene
Hs.22905
Gene Chromosome Location
Chr.X: 131058242-131089885 on Build GRCh38
Transcripts
NM_001282607.1 NM_144967.3 XM_006724729.1 XM_011531280.1

SNP Detail

SNP ID Chromosome Location
rs972229413 Chr.X: 131085578-131085578 on Build GRCh38
rs878971090 Chr.X: 131085619-131085619 on Build GRCh38
rs779502181 Chr.X: 131085643-131085643 on Build GRCh38
rs372255747 Chr.X: 131085644-131085644 on Build GRCh38
rs766007387 Chr.X: 131085584-131085584 on Build GRCh38
rs761692655 Chr.X: 131085562-131085562 on Build GRCh38
rs920782800 Chr.X: 131085588-131085588 on Build GRCh38
rs200086007 Chr.X: 131085592-131085592 on Build GRCh38
rs1015762625 Chr.X: 131085521-131085521 on Build GRCh38
rs754449816 Chr.X: 131085630-131085630 on Build GRCh38
rs367716704 Chr.X: 131085577-131085577 on Build GRCh38
rs764140244 Chr.X: 131085547-131085547 on Build GRCh38
rs911682607 Chr.X: 131085503-131085503 on Build GRCh38
rs1003849906 Chr.X: 131085500-131085500 on Build GRCh38
rs34657540 Chr.X: 131085570-131085571 on Build GRCh38
rs777932916 Chr.X: 131085669-131085669 on Build GRCh38
rs774117962 Chr.X: 131085555-131085555 on Build GRCh38
rs767160538 Chr.X: 131085566-131085566 on Build GRCh38
rs770023095 Chr.X: 131085687-131085687 on Build GRCh38
rs199825373 Chr.X: 131085583-131085583 on Build GRCh38
rs758800583 Chr.X: 131085661-131085661 on Build GRCh38
rs141635498 Chr.X: 131085678-131085678 on Build GRCh38

View Full Product Details