Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 209606233-209606506 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMK1G
Entrez Gene ID
57172
Gene Name
calcium/calmodulin dependent protein kinase IG
Gene Aliases
CLICK3, CLICKIII, VWS1, dJ272L16.1
UniGene
Hs.199068
Gene Chromosome Location
Chr.1: 209583700-209613939 on Build GRCh38
Transcripts
NM_020439.2 XM_017001866.1 XM_017001867.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4668946 c.499C>A p.L167M Chr.1: 209606383-209606383 on Build GRCh38
6840456 c.473C>T p.S158F Chr.1: 209606357-209606357 on Build GRCh38
903284 c.531C>A p.S177S Chr.1: 209606415-209606415 on Build GRCh38
6508581 c.444C>A p.N148K Chr.1: 209606328-209606328 on Build GRCh38
4901492 c.439G>A p.E147K Chr.1: 209606323-209606323 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750895420 Chr.1: 209606285-209606285 on Build GRCh38
rs755367866 Chr.1: 209606308-209606308 on Build GRCh38
rs751801643 Chr.1: 209606275-209606275 on Build GRCh38
rs201992447 Chr.1: 209606462-209606462 on Build GRCh38
rs148684307 Chr.1: 209606439-209606439 on Build GRCh38
rs983665469 Chr.1: 209606403-209606403 on Build GRCh38
rs752297296 Chr.1: 209606436-209606436 on Build GRCh38
rs774446976 Chr.1: 209606351-209606351 on Build GRCh38
rs267598349 Chr.1: 209606400-209606400 on Build GRCh38
rs376936834 Chr.1: 209606293-209606293 on Build GRCh38
rs767522588 Chr.1: 209606366-209606366 on Build GRCh38
rs777272134 Chr.1: 209606322-209606322 on Build GRCh38
rs761856421 Chr.1: 209606282-209606282 on Build GRCh38
rs764337659 Chr.1: 209606272-209606272 on Build GRCh38
rs770903249 Chr.1: 209606329-209606329 on Build GRCh38
rs745333577 Chr.1: 209606392-209606392 on Build GRCh38
rs769577489 Chr.1: 209606473-209606473 on Build GRCh38
rs781164163 Chr.1: 209606466-209606466 on Build GRCh38
rs143406129 Chr.1: 209606343-209606343 on Build GRCh38
rs760818427 Chr.1: 209606387-209606387 on Build GRCh38
rs773291208 Chr.1: 209606379-209606379 on Build GRCh38
rs866051094 Chr.1: 209606319-209606319 on Build GRCh38
rs868513059 Chr.1: 209606444-209606444 on Build GRCh38
rs778778827 Chr.1: 209606316-209606316 on Build GRCh38
rs962736514 Chr.1: 209606363-209606363 on Build GRCh38
rs762023801 Chr.1: 209606364-209606364 on Build GRCh38
rs880783 Chr.1: 209606460-209606460 on Build GRCh38
rs201553423 Chr.1: 209606467-209606467 on Build GRCh38
rs35498535 Chr.1: 209606307-209606307 on Build GRCh38
rs200388906 Chr.1: 209606317-209606317 on Build GRCh38
rs767035299 Chr.1: 209606407-209606407 on Build GRCh38
rs765580376 Chr.1: 209606435-209606435 on Build GRCh38
rs767799311 Chr.1: 209606340-209606340 on Build GRCh38
rs777172542 Chr.1: 209606440-209606440 on Build GRCh38
rs776517584 Chr.1: 209606337-209606337 on Build GRCh38
rs147371769 Chr.1: 209606323-209606323 on Build GRCh38
rs754396153 Chr.1: 209606427-209606427 on Build GRCh38
rs1027660856 Chr.1: 209606331-209606331 on Build GRCh38
rs556793973 Chr.1: 209606305-209606305 on Build GRCh38
rs755455754 Chr.1: 209606430-209606430 on Build GRCh38
rs767572928 Chr.1: 209606284-209606284 on Build GRCh38

View Full Product Details