Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44027343-44027576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF222
Entrez Gene ID
7673
Gene Name
zinc finger protein 222
Gene Aliases
-
UniGene
Hs.279840
Gene Chromosome Location
Chr.19: 44025324-44033110 on Build GRCh38
Transcripts
NM_001129996.1 NM_013360.2 XM_005259208.3 XM_005259209.3 XM_017027237.1 XM_017027238.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
997778 c.193A>G p.T65A Chr.19: 44027421-44027421 on Build GRCh38
3289681 c.227T>C p.V76A Chr.19: 44027455-44027455 on Build GRCh38
7112289 c.245A>T p.Q82L Chr.19: 44027473-44027473 on Build GRCh38
4476184 c.204C>T p.F68F Chr.19: 44027432-44027432 on Build GRCh38
7427017 c.229A>T p.M77L Chr.19: 44027457-44027457 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768862318 Chr.19: 44027478-44027478 on Build GRCh38
rs548146516 Chr.19: 44027447-44027447 on Build GRCh38
rs761941782 Chr.19: 44027490-44027490 on Build GRCh38
rs770363030 Chr.19: 44027510-44027510 on Build GRCh38
rs777203163 Chr.19: 44027437-44027437 on Build GRCh38
rs868416175 Chr.19: 44027411-44027411 on Build GRCh38
rs748941053 Chr.19: 44027438-44027438 on Build GRCh38
rs755640063 Chr.19: 44027434-44027434 on Build GRCh38
rs116222735 Chr.19: 44027462-44027462 on Build GRCh38
rs752594881 Chr.19: 44027416-44027416 on Build GRCh38
rs751744256 Chr.19: 44027463-44027465 on Build GRCh38
rs778612818 Chr.19: 44027448-44027448 on Build GRCh38
rs765429359 Chr.19: 44027520-44027520 on Build GRCh38
rs776919605 Chr.19: 44027481-44027481 on Build GRCh38
rs767255278 Chr.19: 44027538-44027538 on Build GRCh38
rs754782930 Chr.19: 44027402-44027402 on Build GRCh38
rs757414542 Chr.19: 44027473-44027474 on Build GRCh38
rs568138534 Chr.19: 44027455-44027455 on Build GRCh38
rs866081615 Chr.19: 44027549-44027549 on Build GRCh38
rs986917636 Chr.19: 44027459-44027459 on Build GRCh38
rs73556203 Chr.19: 44027550-44027550 on Build GRCh38
rs772449815 Chr.19: 44027467-44027467 on Build GRCh38
rs370000724 Chr.19: 44027470-44027470 on Build GRCh38
rs569741684 Chr.19: 44027531-44027531 on Build GRCh38
rs528770665 Chr.19: 44027401-44027401 on Build GRCh38
rs182791678 Chr.19: 44027518-44027518 on Build GRCh38
rs878873125 Chr.19: 44027380-44027380 on Build GRCh38
rs750630021 Chr.19: 44027522-44027522 on Build GRCh38
rs990719846 Chr.19: 44027405-44027405 on Build GRCh38
rs1038254736 Chr.19: 44027532-44027532 on Build GRCh38

View Full Product Details