Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 95921150-95921377 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERCC6L2
Entrez Gene ID
375748
Gene Name
ERCC excision repair 6 like 2
Gene Aliases
BMFS2, C9orf102, RAD26L, SR278
UniGene
Hs.432364
Gene Chromosome Location
Chr.9: 95875618-96041092 on Build GRCh38
Transcripts
NM_001010895.2 NM_020207.4 XM_011518641.2 XM_011518644.2 XM_011518645.2 XM_011518646.2 XM_011518647.1 XM_011518650.2 XM_017014707.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1111564 c.1255G>A p.D419N Chr.9: 95921238-95921238 on Build GRCh38
7300925 c.1270C>T p.L424F Chr.9: 95921253-95921253 on Build GRCh38
5990745 c.694A>G p.T232A Chr.9: 95921244-95921244 on Build GRCh38
3909259 c.702A>G p.I234M Chr.9: 95921252-95921252 on Build GRCh38
1247002 c.1273C>A p.Q425K Chr.9: 95921256-95921256 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772905545 Chr.9: 95921308-95921308 on Build GRCh38
rs561445484 Chr.9: 95921314-95921314 on Build GRCh38
rs766145218 Chr.9: 95921215-95921215 on Build GRCh38
rs777953933 Chr.9: 95921230-95921230 on Build GRCh38
rs938078498 Chr.9: 95921185-95921185 on Build GRCh38
rs138666484 Chr.9: 95921274-95921274 on Build GRCh38
rs1045749382 Chr.9: 95921280-95921280 on Build GRCh38
rs751512066 Chr.9: 95921220-95921220 on Build GRCh38
rs769171536 Chr.9: 95921288-95921288 on Build GRCh38
rs750116560 Chr.9: 95921184-95921184 on Build GRCh38
rs764954422 Chr.9: 95921179-95921179 on Build GRCh38
rs769252382 Chr.9: 95921306-95921306 on Build GRCh38
rs147361064 Chr.9: 95921240-95921240 on Build GRCh38
rs374484016 Chr.9: 95921312-95921312 on Build GRCh38
rs138517503 Chr.9: 95921263-95921263 on Build GRCh38
rs779194331 Chr.9: 95921255-95921255 on Build GRCh38
rs866414421 Chr.9: 95921309-95921309 on Build GRCh38
rs754997887 Chr.9: 95921228-95921228 on Build GRCh38
rs371003658 Chr.9: 95921241-95921241 on Build GRCh38
rs142685145 Chr.9: 95921211-95921211 on Build GRCh38
rs761315889 Chr.9: 95921297-95921297 on Build GRCh38
rs764007539 Chr.9: 95921234-95921235 on Build GRCh38
rs759491086 Chr.9: 95921344-95921344 on Build GRCh38
rs775003057 Chr.9: 95921311-95921311 on Build GRCh38
rs772464072 Chr.9: 95921267-95921267 on Build GRCh38
rs747471239 Chr.9: 95921285-95921285 on Build GRCh38
rs762803546 Chr.9: 95921213-95921213 on Build GRCh38
rs59765732 Chr.9: 95921268-95921268 on Build GRCh38
rs751422186 Chr.9: 95921328-95921328 on Build GRCh38
rs990785109 Chr.9: 95921222-95921222 on Build GRCh38
rs886037662 Chr.9: 95921219-95921222 on Build GRCh38
rs776058405 Chr.9: 95921294-95921294 on Build GRCh38

View Full Product Details