Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 9072742-9072967 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
A2M
Entrez Gene ID
2
Gene Name
alpha-2-macroglobulin
Gene Aliases
A2MD, CPAMD5, FWP007, S863-7
UniGene
Hs.212838
Gene Chromosome Location
Chr.12: 9067708-9115962 on Build GRCh38
Transcripts
NM_000014.4 XM_006719056.2 XM_017018683.1 XM_017018684.1 XM_017018685.1

Gene Symbol
KLRG1
Entrez Gene ID
10219
Gene Name
killer cell lectin like receptor G1
Gene Aliases
2F1, CLEC15A, MAFA, MAFA-2F1, MAFA-L, MAFA-LIKE
UniGene
Hs.558446
Gene Chromosome Location
Chr.12: 8989079-9255692 on Build GRCh38
Transcripts
NM_000014.4 XM_006719056.2 XM_017018683.1 XM_017018684.1 XM_017018685.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4745963 c.3884A>G p.N1295S Chr.12: 9072744-9072744 on Build GRCh38
4745962 c.3875T>A p.V1292E Chr.12: 9072753-9072753 on Build GRCh38
4774463 c.3797C>A p.A1266E Chr.12: 9072831-9072831 on Build GRCh38
5842037 c.3779C>T p.A1260V Chr.12: 9072849-9072849 on Build GRCh38
3399176 c.3757-1G>T p.? Chr.12: 9072872-9072872 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757663737 Chr.12: 9072907-9072907 on Build GRCh38
rs890471052 Chr.12: 9072790-9072790 on Build GRCh38
rs767161022 Chr.12: 9072823-9072823 on Build GRCh38
rs988722303 Chr.12: 9072903-9072903 on Build GRCh38
rs964870982 Chr.12: 9072805-9072805 on Build GRCh38
rs768091616 Chr.12: 9072912-9072912 on Build GRCh38
rs1033282204 Chr.12: 9072880-9072880 on Build GRCh38
rs771420439 Chr.12: 9072779-9072779 on Build GRCh38
rs949741296 Chr.12: 9072932-9072932 on Build GRCh38
rs777160118 Chr.12: 9072793-9072793 on Build GRCh38
rs376118057 Chr.12: 9072852-9072852 on Build GRCh38
rs773928274 Chr.12: 9072921-9072921 on Build GRCh38
rs773838252 Chr.12: 9072809-9072809 on Build GRCh38
rs768173568 Chr.12: 9072807-9072807 on Build GRCh38
rs747389961 Chr.12: 9072775-9072775 on Build GRCh38
rs766389389 Chr.12: 9072862-9072862 on Build GRCh38
rs979792459 Chr.12: 9072940-9072940 on Build GRCh38
rs370082115 Chr.12: 9072902-9072902 on Build GRCh38
rs746402388 Chr.12: 9072909-9072909 on Build GRCh38
rs747364970 Chr.12: 9072906-9072906 on Build GRCh38
rs773049170 Chr.12: 9072832-9072832 on Build GRCh38
rs892001807 Chr.12: 9072876-9072877 on Build GRCh38
rs773885230 Chr.12: 9072931-9072931 on Build GRCh38
rs781626430 Chr.12: 9072908-9072908 on Build GRCh38
rs761535300 Chr.12: 9072820-9072820 on Build GRCh38
rs372114265 Chr.12: 9072799-9072799 on Build GRCh38
rs758631230 Chr.12: 9072895-9072895 on Build GRCh38
rs752804246 Chr.12: 9072893-9072893 on Build GRCh38
rs755061210 Chr.12: 9072873-9072873 on Build GRCh38
rs778210368 Chr.12: 9072773-9072773 on Build GRCh38
rs765410774 Chr.12: 9072890-9072890 on Build GRCh38
rs947670562 Chr.12: 9072887-9072887 on Build GRCh38

View Full Product Details