Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 122565848-122566099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTX3L
Entrez Gene ID
151636
Gene Name
deltex E3 ubiquitin ligase 3L
Gene Aliases
BBAP
UniGene
Hs.518201
Gene Chromosome Location
Chr.3: 122564338-122575203 on Build GRCh38
Transcripts
NM_138287.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4836394 c.338C>T p.S113F Chr.3: 122566009-122566009 on Build GRCh38
727874 c.312G>C p.L104L Chr.3: 122565983-122565983 on Build GRCh38
1729351 c.289A>C p.T97P Chr.3: 122565960-122565960 on Build GRCh38
202969 c.235G>A p.E79K Chr.3: 122565906-122565906 on Build GRCh38
5720122 c.275C>T p.S92L Chr.3: 122565946-122565946 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761255563 Chr.3: 122565925-122565925 on Build GRCh38
rs754753553 Chr.3: 122566006-122566006 on Build GRCh38
rs747807792 Chr.3: 122566046-122566046 on Build GRCh38
rs201015334 Chr.3: 122565915-122565915 on Build GRCh38
rs757311177 Chr.3: 122566015-122566015 on Build GRCh38
rs753682905 Chr.3: 122566004-122566004 on Build GRCh38
rs938144824 Chr.3: 122565884-122565884 on Build GRCh38
rs760640105 Chr.3: 122565959-122565959 on Build GRCh38
rs749671208 Chr.3: 122566032-122566032 on Build GRCh38
rs147713985 Chr.3: 122565920-122565920 on Build GRCh38
rs368984815 Chr.3: 122566039-122566039 on Build GRCh38
rs201539801 Chr.3: 122566018-122566018 on Build GRCh38
rs751754703 Chr.3: 122566011-122566011 on Build GRCh38
rs775806843 Chr.3: 122565905-122565905 on Build GRCh38
rs990984325 Chr.3: 122565962-122565962 on Build GRCh38
rs780460356 Chr.3: 122566029-122566029 on Build GRCh38
rs756627172 Chr.3: 122566020-122566020 on Build GRCh38
rs1042071159 Chr.3: 122565972-122565972 on Build GRCh38
rs918192895 Chr.3: 122565987-122565987 on Build GRCh38
rs972957461 Chr.3: 122566021-122566021 on Build GRCh38
rs781299398 Chr.3: 122566017-122566017 on Build GRCh38
rs771533413 Chr.3: 122566060-122566060 on Build GRCh38
rs768963281 Chr.3: 122566036-122566036 on Build GRCh38
rs773065070 Chr.3: 122565936-122565936 on Build GRCh38
rs772766142 Chr.3: 122566076-122566076 on Build GRCh38
rs766335984 Chr.3: 122565971-122565971 on Build GRCh38
rs375283808 Chr.3: 122565970-122565970 on Build GRCh38
rs767017007 Chr.3: 122565928-122565928 on Build GRCh38
rs748994219 Chr.3: 122565906-122565906 on Build GRCh38
rs867609206 Chr.3: 122565980-122565980 on Build GRCh38
rs918259593 Chr.3: 122566038-122566038 on Build GRCh38
rs764423884 Chr.3: 122566007-122566007 on Build GRCh38
rs745729800 Chr.3: 122565898-122565900 on Build GRCh38

View Full Product Details