Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 143333225-143333486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIG1
Entrez Gene ID
51390
Gene Name
androgen induced 1
Gene Aliases
AIG-1, dJ95L4.1
UniGene
Hs.567501
Gene Chromosome Location
Chr.6: 143059363-143343594 on Build GRCh38
Transcripts
NM_001286587.1 NM_016108.3 XM_011535881.2 XM_011535882.2 XM_011535883.2 XM_011535884.2 XM_011535885.2 XM_017010927.1 XM_017010928.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3016953 c.525G>T p.W175C Chr.6: 143333291-143333291 on Build GRCh38
6172734 c.674A>T p.Q225L Chr.6: 143333440-143333440 on Build GRCh38
6597199 c.522C>A p.C174* Chr.6: 143333288-143333288 on Build GRCh38
1074042 c.554A>G p.Y185C Chr.6: 143333320-143333320 on Build GRCh38
4350390 c.525G>T p.W175C Chr.6: 143333291-143333291 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770119974 Chr.6: 143333372-143333372 on Build GRCh38
rs781302825 Chr.6: 143333360-143333360 on Build GRCh38
rs112686137 Chr.6: 143333291-143333291 on Build GRCh38
rs780570206 Chr.6: 143333311-143333316 on Build GRCh38
rs780476947 Chr.6: 143333343-143333343 on Build GRCh38
rs994872509 Chr.6: 143333448-143333448 on Build GRCh38
rs759279625 Chr.6: 143333447-143333447 on Build GRCh38
rs769018833 Chr.6: 143333357-143333357 on Build GRCh38
rs758967274 Chr.6: 143333340-143333340 on Build GRCh38
rs761823929 Chr.6: 143333276-143333276 on Build GRCh38
rs377454113 Chr.6: 143333329-143333329 on Build GRCh38
rs766407334 Chr.6: 143333305-143333305 on Build GRCh38
rs769957263 Chr.6: 143333279-143333279 on Build GRCh38
rs768769709 Chr.6: 143333273-143333273 on Build GRCh38
rs749778536 Chr.6: 143333314-143333314 on Build GRCh38
rs140037341 Chr.6: 143333361-143333361 on Build GRCh38
rs773483976 Chr.6: 143333376-143333376 on Build GRCh38
rs911538938 Chr.6: 143333348-143333348 on Build GRCh38
rs747391586 Chr.6: 143333251-143333251 on Build GRCh38
rs775886698 Chr.6: 143333247-143333247 on Build GRCh38
rs767383214 Chr.6: 143333455-143333455 on Build GRCh38
rs372534082 Chr.6: 143333323-143333323 on Build GRCh38
rs777311144 Chr.6: 143333330-143333330 on Build GRCh38
rs774294847 Chr.6: 143333446-143333446 on Build GRCh38
rs753329173 Chr.6: 143333338-143333338 on Build GRCh38
rs987472652 Chr.6: 143333321-143333321 on Build GRCh38
rs371153071 Chr.6: 143333347-143333347 on Build GRCh38
rs141975087 Chr.6: 143333375-143333375 on Build GRCh38
rs751317038 Chr.6: 143333311-143333311 on Build GRCh38
rs763120016 Chr.6: 143333422-143333422 on Build GRCh38
rs373752134 Chr.6: 143333428-143333428 on Build GRCh38
rs763034083 Chr.6: 143333295-143333295 on Build GRCh38
rs767097349 Chr.6: 143333314-143333314 on Build GRCh38
rs759317736 Chr.6: 143333312-143333312 on Build GRCh38
rs773396157 Chr.6: 143333283-143333283 on Build GRCh38
rs112208540 Chr.6: 143333420-143333420 on Build GRCh38

View Full Product Details