Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55822905-55823166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC14
Entrez Gene ID
85406
Gene Name
DnaJ heat shock protein family (Hsp40) member C14
Gene Aliases
DNAJ, DRIP78, HDJ3, LIP6
UniGene
Hs.709320
Gene Chromosome Location
Chr.12: 55820960-55829636 on Build GRCh38
Transcripts
NM_032364.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4043380 c.1602T>C p.T534T Chr.12: 55823102-55823102 on Build GRCh38
7169465 c.1561C>A p.L521M Chr.12: 55823143-55823143 on Build GRCh38
4843719 c.1596G>A p.M532I Chr.12: 55823108-55823108 on Build GRCh38
1195985 c.1579G>A p.D527N Chr.12: 55823125-55823125 on Build GRCh38
7092754 c.1575A>C p.Q525H Chr.12: 55823129-55823129 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372078440 Chr.12: 55823061-55823061 on Build GRCh38
rs150964088 Chr.12: 55823114-55823114 on Build GRCh38
rs747499664 Chr.12: 55823056-55823058 on Build GRCh38
rs562299309 Chr.12: 55823009-55823009 on Build GRCh38
rs540473747 Chr.12: 55822978-55822978 on Build GRCh38
rs768557433 Chr.12: 55823109-55823109 on Build GRCh38
rs144783896 Chr.12: 55823119-55823119 on Build GRCh38
rs751997603 Chr.12: 55823029-55823029 on Build GRCh38
rs182197439 Chr.12: 55823017-55823017 on Build GRCh38
rs931772946 Chr.12: 55822975-55822975 on Build GRCh38
rs746279944 Chr.12: 55823058-55823058 on Build GRCh38
rs1016168922 Chr.12: 55823002-55823002 on Build GRCh38
rs367588622 Chr.12: 55823105-55823105 on Build GRCh38
rs760507468 Chr.12: 55823130-55823130 on Build GRCh38
rs757827203 Chr.12: 55823037-55823037 on Build GRCh38
rs745900085 Chr.12: 55822986-55822986 on Build GRCh38
rs781521133 Chr.12: 55823040-55823040 on Build GRCh38
rs764819292 Chr.12: 55823028-55823028 on Build GRCh38
rs140969766 Chr.12: 55823115-55823115 on Build GRCh38
rs774560051 Chr.12: 55823110-55823110 on Build GRCh38
rs374549407 Chr.12: 55823103-55823103 on Build GRCh38
rs766219836 Chr.12: 55823137-55823137 on Build GRCh38

View Full Product Details