Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 31064784-31065051 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WRN
Entrez Gene ID
7486
Gene Name
Werner syndrome RecQ like helicase
Gene Aliases
RECQ3, RECQL2, RECQL3
UniGene
Hs.632050
Gene Chromosome Location
Chr.8: 31033262-31173761 on Build GRCh38
Transcripts
NM_000553.4 XM_011544639.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6327233 c.356-7G>T p.? Chr.8: 31064908-31064908 on Build GRCh38
6430149 c.365A>T p.Q122L Chr.8: 31064924-31064924 on Build GRCh38
5611175 c.406G>T p.A136S Chr.8: 31064965-31064965 on Build GRCh38
1099219 c.460A>G p.K154E Chr.8: 31065019-31065019 on Build GRCh38
6253918 c.412G>T p.V138L Chr.8: 31064971-31064971 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs267607008 Chr.8: 31064962-31064962 on Build GRCh38
rs772202608 Chr.8: 31064907-31064907 on Build GRCh38
rs768174465 Chr.8: 31064921-31064921 on Build GRCh38
rs993699533 Chr.8: 31064819-31064819 on Build GRCh38
rs780068269 Chr.8: 31064913-31064913 on Build GRCh38
rs267607007 Chr.8: 31064932-31064932 on Build GRCh38
rs387906337 Chr.8: 31064934-31064934 on Build GRCh38
rs745644260 Chr.8: 31064892-31064892 on Build GRCh38
rs925567153 Chr.8: 31065003-31065003 on Build GRCh38
rs767673195 Chr.8: 31065001-31065001 on Build GRCh38
rs754996830 Chr.8: 31064877-31064877 on Build GRCh38
rs587778753 Chr.8: 31065002-31065002 on Build GRCh38
rs746528374 Chr.8: 31064914-31064914 on Build GRCh38
rs752921218 Chr.8: 31064886-31064886 on Build GRCh38
rs773697397 Chr.8: 31064988-31064988 on Build GRCh38
rs766887542 Chr.8: 31064991-31064991 on Build GRCh38
rs548061402 Chr.8: 31064979-31064979 on Build GRCh38
rs61761625 Chr.8: 31065005-31065005 on Build GRCh38
rs752693894 Chr.8: 31065004-31065004 on Build GRCh38
rs867641943 Chr.8: 31064954-31064954 on Build GRCh38
rs778813155 Chr.8: 31064891-31064891 on Build GRCh38
rs17847582 Chr.8: 31064965-31064965 on Build GRCh38
rs752014453 Chr.8: 31064995-31064995 on Build GRCh38
rs751646028 Chr.8: 31064870-31064870 on Build GRCh38
rs184973448 Chr.8: 31064869-31064869 on Build GRCh38
rs2725346 Chr.8: 31064830-31064830 on Build GRCh38
rs771444804 Chr.8: 31064990-31064990 on Build GRCh38
rs372942234 Chr.8: 31064922-31064922 on Build GRCh38
rs566780727 Chr.8: 31064890-31064890 on Build GRCh38
rs200456571 Chr.8: 31065011-31065011 on Build GRCh38
rs951969651 Chr.8: 31064953-31064953 on Build GRCh38
rs769317785 Chr.8: 31064980-31064980 on Build GRCh38
rs369976838 Chr.8: 31064895-31064895 on Build GRCh38
rs986860495 Chr.8: 31064845-31064845 on Build GRCh38
rs781294554 Chr.8: 31064883-31064883 on Build GRCh38
rs878854139 Chr.8: 31064989-31064989 on Build GRCh38
rs971264760 Chr.8: 31064818-31064818 on Build GRCh38
rs916722009 Chr.8: 31065016-31065016 on Build GRCh38
rs1028146399 Chr.8: 31064826-31064826 on Build GRCh38

View Full Product Details