Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100737149-100737410 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZAN
Entrez Gene ID
7455
Gene Name
zonadhesin (gene/pseudogene)
Gene Aliases
-
UniGene
Hs.307004
Gene Chromosome Location
Chr.7: 100733599-100797797 on Build GRCh38
Transcripts
NM_003386.2 NM_173059.2 NR_111917.1 NR_111918.1 NR_111919.1 XM_011516555.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1082965 c.571G>A p.A191T Chr.7: 100737307-100737307 on Build GRCh38
7244260 c.596G>A p.R199Q Chr.7: 100737332-100737332 on Build GRCh38
278179 c.611G>A p.R204H Chr.7: 100737347-100737347 on Build GRCh38
4977616 c.612C>T p.R204R Chr.7: 100737348-100737348 on Build GRCh38
5406015 c.570C>T p.I190I Chr.7: 100737306-100737306 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs267601184 Chr.7: 100737346-100737346 on Build GRCh38
rs1042133847 Chr.7: 100737385-100737385 on Build GRCh38
rs746635287 Chr.7: 100737327-100737327 on Build GRCh38
rs778724541 Chr.7: 100737297-100737297 on Build GRCh38
rs753605191 Chr.7: 100737351-100737351 on Build GRCh38
rs755013926 Chr.7: 100737268-100737268 on Build GRCh38
rs760143173 Chr.7: 100737336-100737336 on Build GRCh38
rs780711175 Chr.7: 100737386-100737386 on Build GRCh38
rs114183970 Chr.7: 100737280-100737280 on Build GRCh38
rs757017365 Chr.7: 100737359-100737359 on Build GRCh38
rs771401230 Chr.7: 100737332-100737332 on Build GRCh38
rs961239051 Chr.7: 100737210-100737210 on Build GRCh38
rs183887733 Chr.7: 100737282-100737282 on Build GRCh38
rs761520906 Chr.7: 100737329-100737329 on Build GRCh38
rs763258626 Chr.7: 100737331-100737331 on Build GRCh38
rs528229272 Chr.7: 100737295-100737295 on Build GRCh38
rs774730226 Chr.7: 100737334-100737334 on Build GRCh38
rs367593224 Chr.7: 100737347-100737347 on Build GRCh38
rs750324983 Chr.7: 100737371-100737371 on Build GRCh38
rs749062113 Chr.7: 100737294-100737294 on Build GRCh38
rs773574912 Chr.7: 100737329-100737329 on Build GRCh38
rs530702519 Chr.7: 100737209-100737209 on Build GRCh38
rs777692278 Chr.7: 100737293-100737293 on Build GRCh38
rs752952727 Chr.7: 100737281-100737281 on Build GRCh38
rs773605916 Chr.7: 100737228-100737228 on Build GRCh38
rs746543240 Chr.7: 100737382-100737382 on Build GRCh38
rs768345294 Chr.7: 100737328-100737328 on Build GRCh38
rs766817422 Chr.7: 100737236-100737236 on Build GRCh38
rs375717396 Chr.7: 100737381-100737381 on Build GRCh38
rs371861902 Chr.7: 100737348-100737348 on Build GRCh38
rs763492423 Chr.7: 100737231-100737231 on Build GRCh38
rs767196132 Chr.7: 100737373-100737373 on Build GRCh38
rs778827164 Chr.7: 100737369-100737369 on Build GRCh38
rs992743007 Chr.7: 100737211-100737211 on Build GRCh38
rs1015388678 Chr.7: 100737208-100737208 on Build GRCh38
rs1024922051 Chr.7: 100737220-100737220 on Build GRCh38
rs10243399 Chr.7: 100737232-100737232 on Build GRCh38
rs751617675 Chr.7: 100737249-100737249 on Build GRCh38
rs374091129 Chr.7: 100737306-100737306 on Build GRCh38
rs562932336 Chr.7: 100737184-100737184 on Build GRCh38
rs866940196 Chr.7: 100737263-100737263 on Build GRCh38
rs910794533 Chr.7: 100737173-100737173 on Build GRCh38

View Full Product Details