Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 49717608-49717833 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM117A
Entrez Gene ID
81558
Gene Name
family with sequence similarity 117 member A
Gene Aliases
-
UniGene
Hs.514308
Gene Chromosome Location
Chr.17: 49710325-49764156 on Build GRCh38
Transcripts
NM_030802.3 XM_017025182.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3518939 c.812C>T p.P271L Chr.17: 49717611-49717611 on Build GRCh38
6350924 c.733C>T p.R245W Chr.17: 49717690-49717690 on Build GRCh38
4842392 c.709-1G>A p.? Chr.17: 49717715-49717715 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761089718 Chr.17: 49717750-49717750 on Build GRCh38
rs756476334 Chr.17: 49717669-49717669 on Build GRCh38
rs780753655 Chr.17: 49717704-49717704 on Build GRCh38
rs201150618 Chr.17: 49717729-49717729 on Build GRCh38
rs746599310 Chr.17: 49717697-49717697 on Build GRCh38
rs369414429 Chr.17: 49717730-49717730 on Build GRCh38
rs35495960 Chr.17: 49717673-49717673 on Build GRCh38
rs990057749 Chr.17: 49717793-49717793 on Build GRCh38
rs923092098 Chr.17: 49717727-49717727 on Build GRCh38
rs767727058 Chr.17: 49717650-49717650 on Build GRCh38
rs757926166 Chr.17: 49717682-49717682 on Build GRCh38
rs754334415 Chr.17: 49717761-49717761 on Build GRCh38
rs1044387826 Chr.17: 49717786-49717786 on Build GRCh38
rs749236248 Chr.17: 49717723-49717723 on Build GRCh38
rs759834758 Chr.17: 49717646-49717646 on Build GRCh38
rs549856010 Chr.17: 49717659-49717659 on Build GRCh38
rs754676704 Chr.17: 49717699-49717699 on Build GRCh38
rs140500484 Chr.17: 49717690-49717690 on Build GRCh38
rs374371529 Chr.17: 49717755-49717755 on Build GRCh38
rs970733196 Chr.17: 49717705-49717705 on Build GRCh38
rs113987142 Chr.17: 49717716-49717716 on Build GRCh38
rs570857941 Chr.17: 49717783-49717783 on Build GRCh38
rs759586119 Chr.17: 49717738-49717738 on Build GRCh38
rs769642885 Chr.17: 49717720-49717720 on Build GRCh38
rs775644452 Chr.17: 49717747-49717747 on Build GRCh38
rs753084610 Chr.17: 49717661-49717661 on Build GRCh38
rs774498092 Chr.17: 49717642-49717642 on Build GRCh38
rs905974509 Chr.17: 49717800-49717800 on Build GRCh38
rs368778142 Chr.17: 49717719-49717719 on Build GRCh38
rs117876393 Chr.17: 49717722-49717722 on Build GRCh38
rs1027541019 Chr.17: 49717648-49717648 on Build GRCh38
rs34407795 Chr.17: 49717719-49717720 on Build GRCh38

View Full Product Details