Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 69137694-69137967 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UTP4
Entrez Gene ID
84916
Gene Name
UTP4, small subunit processome component
Gene Aliases
CIRH1A, CIRHIN, NAIC, TEX292
UniGene
Hs.461113
Gene Chromosome Location
Chr.16: 69132596-69169034 on Build GRCh38
Transcripts
NM_001318391.1 NM_032830.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4571147 c.389T>A p.I130N Chr.16: 69137838-69137838 on Build GRCh38
349234 c.431A>T p.Q144L Chr.16: 69137880-69137880 on Build GRCh38
4062216 c.425A>G p.D142G Chr.16: 69137874-69137874 on Build GRCh38
6371789 c.352G>T p.V118F Chr.16: 69137801-69137801 on Build GRCh38
1201216 c.356G>T p.G119V Chr.16: 69137805-69137805 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1039544664 Chr.16: 69137728-69137728 on Build GRCh38
rs746904135 Chr.16: 69137923-69137923 on Build GRCh38
rs758138485 Chr.16: 69137768-69137769 on Build GRCh38
rs202164680 Chr.16: 69137891-69137891 on Build GRCh38
rs371231272 Chr.16: 69137937-69137937 on Build GRCh38
rs773201079 Chr.16: 69137814-69137814 on Build GRCh38
rs772044972 Chr.16: 69137797-69137797 on Build GRCh38
rs374368864 Chr.16: 69137750-69137750 on Build GRCh38
rs748042307 Chr.16: 69137792-69137792 on Build GRCh38
rs1037834445 Chr.16: 69137775-69137775 on Build GRCh38
rs368415223 Chr.16: 69137785-69137785 on Build GRCh38
rs746046263 Chr.16: 69137783-69137783 on Build GRCh38
rs926292225 Chr.16: 69137720-69137720 on Build GRCh38
rs572699127 Chr.16: 69137764-69137764 on Build GRCh38
rs184778917 Chr.16: 69137724-69137724 on Build GRCh38
rs781484069 Chr.16: 69137895-69137895 on Build GRCh38
rs1011990327 Chr.16: 69137786-69137786 on Build GRCh38
rs897874271 Chr.16: 69137802-69137802 on Build GRCh38
rs879125546 Chr.16: 69137757-69137757 on Build GRCh38
rs369049138 Chr.16: 69137894-69137894 on Build GRCh38
rs562350763 Chr.16: 69137917-69137917 on Build GRCh38
rs200047779 Chr.16: 69137876-69137876 on Build GRCh38
rs777821483 Chr.16: 69137920-69137920 on Build GRCh38
rs915076599 Chr.16: 69137727-69137728 on Build GRCh38
rs764657942 Chr.16: 69137880-69137880 on Build GRCh38
rs550525664 Chr.16: 69137909-69137909 on Build GRCh38
rs532322915 Chr.16: 69137908-69137908 on Build GRCh38
rs758885251 Chr.16: 69137885-69137885 on Build GRCh38
rs766174741 Chr.16: 69137836-69137836 on Build GRCh38
rs781357188 Chr.16: 69137774-69137774 on Build GRCh38
rs3217261 Chr.16: 69137728-69137729 on Build GRCh38
rs765883944 Chr.16: 69137786-69137787 on Build GRCh38
rs376840053 Chr.16: 69137892-69137892 on Build GRCh38
rs770862010 Chr.16: 69137932-69137932 on Build GRCh38
rs752244922 Chr.16: 69137883-69137883 on Build GRCh38
rs776455835 Chr.16: 69137873-69137873 on Build GRCh38
rs750661522 Chr.16: 69137904-69137904 on Build GRCh38
rs749667961 Chr.16: 69137729-69137729 on Build GRCh38
rs778064509 Chr.16: 69137759-69137759 on Build GRCh38
rs1030260329 Chr.16: 69137905-69137905 on Build GRCh38
rs776502169 Chr.16: 69137935-69137935 on Build GRCh38
rs989237106 Chr.16: 69137723-69137725 on Build GRCh38
rs866919333 Chr.16: 69137842-69137842 on Build GRCh38
rs1048814262 Chr.16: 69137727-69137727 on Build GRCh38
rs751152945 Chr.16: 69137868-69137869 on Build GRCh38
rs760526066 Chr.16: 69137828-69137828 on Build GRCh38
rs541930560 Chr.16: 69137748-69137748 on Build GRCh38
rs933342896 Chr.16: 69137734-69137734 on Build GRCh38
rs775586511 Chr.16: 69137789-69137789 on Build GRCh38
rs1003652868 Chr.16: 69137840-69137840 on Build GRCh38

View Full Product Details