Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 49692706-49692973 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRISP2
Entrez Gene ID
7180
Gene Name
cysteine rich secretory protein 2
Gene Aliases
CRISP-2, CT36, GAPDL5, TPX1, TSP1
UniGene
Hs.2042
Gene Chromosome Location
Chr.6: 49676665-49714198 on Build GRCh38
Transcripts
NM_001142407.2 NM_001142408.2 NM_001142417.2 NM_001142435.2 NM_001261822.1 NM_003296.3 XM_005249349.1 XM_005249350.1 XM_005249351.3 XM_005249352.3 XM_005249353.3 XM_005249356.1 XM_011514841.1 XM_011514842.1 XM_011514843.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1080079 c.628C>A p.L210I Chr.6: 49692877-49692877 on Build GRCh38
4503794 c.648C>T p.S216S Chr.6: 49692857-49692857 on Build GRCh38
1445009 c.701C>A p.A234D Chr.6: 49692804-49692804 on Build GRCh38
6477531 c.697A>C p.K233Q Chr.6: 49692808-49692808 on Build GRCh38
3394237 c.605-7G>C p.? Chr.6: 49692907-49692907 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764283148 Chr.6: 49692945-49692945 on Build GRCh38
rs780653237 Chr.6: 49692883-49692883 on Build GRCh38
rs759279881 Chr.6: 49692931-49692931 on Build GRCh38
rs749710327 Chr.6: 49692741-49692741 on Build GRCh38
rs1049784604 Chr.6: 49692824-49692824 on Build GRCh38
rs772587233 Chr.6: 49692894-49692894 on Build GRCh38
rs930882229 Chr.6: 49692805-49692805 on Build GRCh38
rs558511264 Chr.6: 49692754-49692754 on Build GRCh38
rs762512665 Chr.6: 49692783-49692783 on Build GRCh38
rs972280399 Chr.6: 49692740-49692740 on Build GRCh38
rs772726407 Chr.6: 49692875-49692875 on Build GRCh38
rs779371926 Chr.6: 49692865-49692865 on Build GRCh38
rs919435094 Chr.6: 49692760-49692760 on Build GRCh38
rs577428520 Chr.6: 49692782-49692782 on Build GRCh38
rs757394764 Chr.6: 49692864-49692864 on Build GRCh38
rs772671444 Chr.6: 49692733-49692733 on Build GRCh38
rs761354474 Chr.6: 49692735-49692735 on Build GRCh38
rs747668694 Chr.6: 49692885-49692885 on Build GRCh38
rs759511323 Chr.6: 49692795-49692795 on Build GRCh38
rs544121328 Chr.6: 49692812-49692812 on Build GRCh38
rs776309599 Chr.6: 49692734-49692734 on Build GRCh38
rs369082725 Chr.6: 49692914-49692914 on Build GRCh38
rs564116939 Chr.6: 49692835-49692835 on Build GRCh38
rs760592389 Chr.6: 49692934-49692934 on Build GRCh38
rs910514075 Chr.6: 49692866-49692866 on Build GRCh38
rs767404432 Chr.6: 49692804-49692804 on Build GRCh38
rs562593669 Chr.6: 49692935-49692935 on Build GRCh38
rs777709973 Chr.6: 49692847-49692847 on Build GRCh38
rs34093486 Chr.6: 49692816-49692817 on Build GRCh38
rs754147443 Chr.6: 49692849-49692849 on Build GRCh38
rs751290579 Chr.6: 49692787-49692787 on Build GRCh38
rs766258708 Chr.6: 49692785-49692785 on Build GRCh38
rs778638236 Chr.6: 49692764-49692765 on Build GRCh38
rs767445386 Chr.6: 49692933-49692933 on Build GRCh38
rs76267147 Chr.6: 49692762-49692762 on Build GRCh38
rs774235367 Chr.6: 49692915-49692915 on Build GRCh38
rs762716256 Chr.6: 49692913-49692913 on Build GRCh38
rs746176594 Chr.6: 49692868-49692868 on Build GRCh38
rs756118867 Chr.6: 49692843-49692843 on Build GRCh38
rs769500059 Chr.6: 49692889-49692889 on Build GRCh38

View Full Product Details