Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 184590600-184590851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf21
Entrez Gene ID
81563
Gene Name
chromosome 1 open reading frame 21
Gene Aliases
PIG13
UniGene
Hs.497159
Gene Chromosome Location
Chr.1: 184387016-184629021 on Build GRCh38
Transcripts
NM_030806.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5993385 c.226A>T p.N76Y Chr.1: 184590775-184590775 on Build GRCh38
6615226 c.89A>G p.E30G Chr.1: 184590740-184590740 on Build GRCh38
6615225 c.191A>G p.E64G Chr.1: 184590740-184590740 on Build GRCh38
4143116 c.198T>G p.S66R Chr.1: 184590747-184590747 on Build GRCh38
5993386 c.124A>T p.N42Y Chr.1: 184590775-184590775 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372714212 Chr.1: 184590713-184590713 on Build GRCh38
rs745820026 Chr.1: 184590784-184590784 on Build GRCh38
rs747903910 Chr.1: 184590647-184590647 on Build GRCh38
rs765431561 Chr.1: 184590762-184590762 on Build GRCh38
rs541123924 Chr.1: 184590798-184590798 on Build GRCh38
rs779620403 Chr.1: 184590789-184590789 on Build GRCh38
rs780698724 Chr.1: 184590623-184590623 on Build GRCh38
rs755627276 Chr.1: 184590788-184590788 on Build GRCh38
rs766471630 Chr.1: 184590728-184590728 on Build GRCh38
rs755024048 Chr.1: 184590641-184590641 on Build GRCh38
rs867556439 Chr.1: 184590766-184590766 on Build GRCh38
rs550345064 Chr.1: 184590707-184590707 on Build GRCh38
rs114750800 Chr.1: 184590628-184590628 on Build GRCh38
rs781232394 Chr.1: 184590780-184590780 on Build GRCh38
rs768835490 Chr.1: 184590695-184590695 on Build GRCh38
rs369125601 Chr.1: 184590703-184590703 on Build GRCh38
rs773751031 Chr.1: 184590714-184590714 on Build GRCh38
rs755237546 Chr.1: 184590748-184590748 on Build GRCh38
rs753953314 Chr.1: 184590744-184590744 on Build GRCh38
rs149800099 Chr.1: 184590812-184590812 on Build GRCh38
rs12754557 Chr.1: 184590654-184590654 on Build GRCh38
rs771822974 Chr.1: 184590666-184590666 on Build GRCh38
rs571876747 Chr.1: 184590633-184590633 on Build GRCh38
rs112175521 Chr.1: 184590816-184590816 on Build GRCh38
rs760708785 Chr.1: 184590727-184590727 on Build GRCh38
rs1051025007 Chr.1: 184590626-184590626 on Build GRCh38
rs148848494 Chr.1: 184590629-184590629 on Build GRCh38
rs980068778 Chr.1: 184590674-184590674 on Build GRCh38
rs751343261 Chr.1: 184590769-184590769 on Build GRCh38
rs1013048345 Chr.1: 184590656-184590656 on Build GRCh38

View Full Product Details