Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33316768-33317015 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZBTB22
Entrez Gene ID
9278
Gene Name
zinc finger and BTB domain containing 22
Gene Aliases
BING1, ZBTB22A, ZNF297, ZNF297A, fru, fruitless
UniGene
Hs.206770
Gene Chromosome Location
Chr.6: 33314405-33317942 on Build GRCh38
Transcripts
NM_001145338.1 NM_005453.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7423807 c.122G>A p.S41N Chr.6: 33316795-33316795 on Build GRCh38
5194684 c.31delG p.A11fs*29 Chr.6: 33316886-33316886 on Build GRCh38
6578454 c.34G>A p.A12T Chr.6: 33316883-33316883 on Build GRCh38
3994880 c.26G>A p.S9N Chr.6: 33316891-33316891 on Build GRCh38
6442236 c.109C>T p.P37S Chr.6: 33316808-33316808 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368120208 Chr.6: 33316982-33316983 on Build GRCh38
rs35958299 Chr.6: 33316928-33316929 on Build GRCh38
rs776281920 Chr.6: 33316938-33316938 on Build GRCh38
rs184607829 Chr.6: 33316971-33316971 on Build GRCh38
rs563554175 Chr.6: 33316802-33316802 on Build GRCh38
rs763407369 Chr.6: 33316934-33316935 on Build GRCh38
rs376554920 Chr.6: 33316890-33316890 on Build GRCh38
rs747185754 Chr.6: 33316880-33316880 on Build GRCh38
rs1011650765 Chr.6: 33316922-33316922 on Build GRCh38
rs769118531 Chr.6: 33316935-33316936 on Build GRCh38
rs917633079 Chr.6: 33316937-33316937 on Build GRCh38
rs141192416 Chr.6: 33316854-33316854 on Build GRCh38
rs776477326 Chr.6: 33316838-33316838 on Build GRCh38
rs192756182 Chr.6: 33316951-33316951 on Build GRCh38
rs766700691 Chr.6: 33316870-33316870 on Build GRCh38
rs575402967 Chr.6: 33316850-33316850 on Build GRCh38
rs759314951 Chr.6: 33316844-33316844 on Build GRCh38
rs770541314 Chr.6: 33316836-33316836 on Build GRCh38
rs772733259 Chr.6: 33316819-33316819 on Build GRCh38
rs758921712 Chr.6: 33316921-33316921 on Build GRCh38
rs752936758 Chr.6: 33316889-33316889 on Build GRCh38
rs755404727 Chr.6: 33316892-33316892 on Build GRCh38
rs774251980 Chr.6: 33316848-33316848 on Build GRCh38
rs779271056 Chr.6: 33316798-33316798 on Build GRCh38
rs961638471 Chr.6: 33316958-33316958 on Build GRCh38
rs750493100 Chr.6: 33316856-33316856 on Build GRCh38
rs953027119 Chr.6: 33316981-33316981 on Build GRCh38
rs199691306 Chr.6: 33316876-33316876 on Build GRCh38
rs746518897 Chr.6: 33316932-33316932 on Build GRCh38
rs762906555 Chr.6: 33316851-33316851 on Build GRCh38
rs148229343 Chr.6: 33316800-33316800 on Build GRCh38
rs1015116929 Chr.6: 33316812-33316812 on Build GRCh38
rs1055311217 Chr.6: 33316821-33316821 on Build GRCh38
rs1056781237 Chr.6: 33316878-33316878 on Build GRCh38
rs79989637 Chr.6: 33316952-33316952 on Build GRCh38
rs777192990 Chr.6: 33316928-33316928 on Build GRCh38
rs372107453 Chr.6: 33316935-33316935 on Build GRCh38
rs755606636 Chr.6: 33316795-33316795 on Build GRCh38
rs746717070 Chr.6: 33316823-33316823 on Build GRCh38
rs990449216 Chr.6: 33316936-33316936 on Build GRCh38
rs756440751 Chr.6: 33316869-33316869 on Build GRCh38
rs1028862790 Chr.6: 33316934-33316934 on Build GRCh38
rs201803016 Chr.6: 33316975-33316976 on Build GRCh38
rs748628590 Chr.6: 33316917-33316917 on Build GRCh38
rs373933633 Chr.6: 33316966-33316966 on Build GRCh38
rs1053568637 Chr.6: 33316908-33316908 on Build GRCh38

View Full Product Details