Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 13318815-13319082 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN3L
Entrez Gene ID
92552
Gene Name
ataxin 3 like
Gene Aliases
MJDL
UniGene
Hs.382641
Gene Chromosome Location
Chr.X: 13318647-13320399 on Build GRCh38
Transcripts
NM_001135995.1 NR_046087.1

Gene Symbol
GS1-600G8.3
Entrez Gene ID
100093698
Gene Name
unknown transcript
Gene Aliases
-
UniGene
Hs.643612
Gene Chromosome Location
Chr.X: 13310652-13319933 on Build GRCh38
Transcripts
NM_001135995.1 NR_046087.1

SNP Detail

SNP ID Chromosome Location
rs200821460 Chr.X: 13318909-13318909 on Build GRCh38
rs751991266 Chr.X: 13319027-13319027 on Build GRCh38
rs978675466 Chr.X: 13318919-13318919 on Build GRCh38
rs780131230 Chr.X: 13319015-13319015 on Build GRCh38
rs765392102 Chr.X: 13319003-13319003 on Build GRCh38
rs199968559 Chr.X: 13318932-13318932 on Build GRCh38
rs373799323 Chr.X: 13319002-13319002 on Build GRCh38
rs766819464 Chr.X: 13318898-13318898 on Build GRCh38
rs769321240 Chr.X: 13318982-13318982 on Build GRCh38
rs922396528 Chr.X: 13318948-13318948 on Build GRCh38
rs754028227 Chr.X: 13318901-13318901 on Build GRCh38
rs947834910 Chr.X: 13319042-13319044 on Build GRCh38
rs748673522 Chr.X: 13319037-13319037 on Build GRCh38
rs777339561 Chr.X: 13319046-13319046 on Build GRCh38
rs187826331 Chr.X: 13318921-13318921 on Build GRCh38
rs750682579 Chr.X: 13318926-13318926 on Build GRCh38
rs376950658 Chr.X: 13318927-13318927 on Build GRCh38
rs201988976 Chr.X: 13318973-13318973 on Build GRCh38
rs368171709 Chr.X: 13318890-13318890 on Build GRCh38
rs4830842 Chr.X: 13318940-13318940 on Build GRCh38
rs755301446 Chr.X: 13319031-13319031 on Build GRCh38
rs377086256 Chr.X: 13318846-13318846 on Build GRCh38
rs369915965 Chr.X: 13319001-13319001 on Build GRCh38
rs192647842 Chr.X: 13319025-13319025 on Build GRCh38
rs756507822 Chr.X: 13318897-13318901 on Build GRCh38
rs748686220 Chr.X: 13319050-13319050 on Build GRCh38
rs772376884 Chr.X: 13318986-13318988 on Build GRCh38
rs751358610 Chr.X: 13318876-13318876 on Build GRCh38
rs769214229 Chr.X: 13318935-13318935 on Build GRCh38
rs770707973 Chr.X: 13319033-13319033 on Build GRCh38
rs758662068 Chr.X: 13319006-13319006 on Build GRCh38
rs750406543 Chr.X: 13319004-13319004 on Build GRCh38
rs770421590 Chr.X: 13318991-13318991 on Build GRCh38
rs779303424 Chr.X: 13318935-13318936 on Build GRCh38
rs17322143 Chr.X: 13318996-13318996 on Build GRCh38
rs376418054 Chr.X: 13319044-13319044 on Build GRCh38
rs991638578 Chr.X: 13318870-13318870 on Build GRCh38

View Full Product Details