Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 72443554-72443815 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF366
Entrez Gene ID
167465
Gene Name
zinc finger protein 366
Gene Aliases
DCSCRIPT
UniGene
Hs.224794
Gene Chromosome Location
Chr.5: 72439899-72507422 on Build GRCh38
Transcripts
NM_152625.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3429614 c.2185G>T p.E729* Chr.5: 72443806-72443806 on Build GRCh38
6304627 c.2194G>T p.E732* Chr.5: 72443797-72443797 on Build GRCh38
4535486 c.2198G>A p.R733K Chr.5: 72443793-72443793 on Build GRCh38
5904818 c.2216C>G p.A739G Chr.5: 72443775-72443775 on Build GRCh38
6443286 c.2206G>A p.E736K Chr.5: 72443785-72443785 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199641622 Chr.5: 72443780-72443780 on Build GRCh38
rs770111919 Chr.5: 72443709-72443709 on Build GRCh38
rs374173373 Chr.5: 72443713-72443713 on Build GRCh38
rs1041160963 Chr.5: 72443773-72443773 on Build GRCh38
rs920221002 Chr.5: 72443676-72443676 on Build GRCh38
rs4267851 Chr.5: 72443719-72443719 on Build GRCh38
rs764871895 Chr.5: 72443726-72443726 on Build GRCh38
rs1023334731 Chr.5: 72443765-72443765 on Build GRCh38
rs750134552 Chr.5: 72443760-72443760 on Build GRCh38
rs879071676 Chr.5: 72443668-72443668 on Build GRCh38
rs758115238 Chr.5: 72443763-72443763 on Build GRCh38
rs757290919 Chr.5: 72443706-72443706 on Build GRCh38
rs760117737 Chr.5: 72443750-72443750 on Build GRCh38
rs776294439 Chr.5: 72443762-72443762 on Build GRCh38
rs1050495865 Chr.5: 72443631-72443631 on Build GRCh38
rs565290190 Chr.5: 72443666-72443666 on Build GRCh38
rs747944618 Chr.5: 72443716-72443716 on Build GRCh38
rs757072499 Chr.5: 72443665-72443665 on Build GRCh38
rs13188519 Chr.5: 72443775-72443775 on Build GRCh38
rs139079914 Chr.5: 72443628-72443628 on Build GRCh38
rs752044173 Chr.5: 72443777-72443777 on Build GRCh38
rs190965994 Chr.5: 72443577-72443577 on Build GRCh38
rs764846541 Chr.5: 72443764-72443764 on Build GRCh38
rs371219891 Chr.5: 72443707-72443707 on Build GRCh38
rs542926512 Chr.5: 72443703-72443703 on Build GRCh38
rs557112810 Chr.5: 72443654-72443654 on Build GRCh38
rs770424180 Chr.5: 72443751-72443751 on Build GRCh38
rs866568398 Chr.5: 72443608-72443608 on Build GRCh38
rs77724139 Chr.5: 72443692-72443692 on Build GRCh38
rs961951613 Chr.5: 72443616-72443616 on Build GRCh38
rs577762280 Chr.5: 72443620-72443620 on Build GRCh38
rs987974090 Chr.5: 72443579-72443579 on Build GRCh38
rs750512108 Chr.5: 72443785-72443785 on Build GRCh38
rs781375221 Chr.5: 72443784-72443784 on Build GRCh38
rs970458957 Chr.5: 72443613-72443613 on Build GRCh38
rs753114079 Chr.5: 72443771-72443771 on Build GRCh38
rs923240613 Chr.5: 72443627-72443627 on Build GRCh38

View Full Product Details