Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 46177829-46178080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH22
Entrez Gene ID
64405
Gene Name
cadherin 22
Gene Aliases
C20orf25, dJ998H6.1
UniGene
Hs.472861
Gene Chromosome Location
Chr.20: 46173733-46308498 on Build GRCh38
Transcripts
NM_021248.2 XM_011528994.2 XM_011528995.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6351813 c.1872C>T p.P624P Chr.20: 46177989-46177989 on Build GRCh38
6624828 c.1874G>A p.G625D Chr.20: 46177987-46177987 on Build GRCh38
3991747 c.1858G>A p.A620T Chr.20: 46178003-46178003 on Build GRCh38
6841906 c.1781C>T p.T594M Chr.20: 46178080-46178080 on Build GRCh38
7103555 c.1879C>A p.L627I Chr.20: 46177982-46177982 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372950957 Chr.20: 46178049-46178049 on Build GRCh38
rs781500013 Chr.20: 46177950-46177950 on Build GRCh38
rs762288213 Chr.20: 46177924-46177924 on Build GRCh38
rs778679068 Chr.20: 46178005-46178005 on Build GRCh38
rs765796879 Chr.20: 46177934-46177934 on Build GRCh38
rs751544343 Chr.20: 46177992-46177992 on Build GRCh38
rs73598378 Chr.20: 46177989-46177989 on Build GRCh38
rs747788571 Chr.20: 46178046-46178046 on Build GRCh38
rs370838347 Chr.20: 46178020-46178020 on Build GRCh38
rs34603854 Chr.20: 46178004-46178004 on Build GRCh38
rs778732199 Chr.20: 46177968-46177968 on Build GRCh38
rs773645188 Chr.20: 46178054-46178054 on Build GRCh38
rs899588457 Chr.20: 46177908-46177908 on Build GRCh38
rs750227357 Chr.20: 46178008-46178008 on Build GRCh38
rs377315839 Chr.20: 46177896-46177896 on Build GRCh38
rs768178720 Chr.20: 46178042-46178042 on Build GRCh38
rs35412957 Chr.20: 46177988-46177988 on Build GRCh38
rs886973029 Chr.20: 46177879-46177879 on Build GRCh38
rs745795688 Chr.20: 46177971-46177971 on Build GRCh38
rs144477780 Chr.20: 46178003-46178003 on Build GRCh38
rs761595615 Chr.20: 46177986-46177986 on Build GRCh38
rs370997653 Chr.20: 46177923-46177923 on Build GRCh38
rs776798469 Chr.20: 46177911-46177911 on Build GRCh38
rs376069500 Chr.20: 46177962-46177962 on Build GRCh38
rs763398311 Chr.20: 46178055-46178055 on Build GRCh38
rs148863832 Chr.20: 46178019-46178019 on Build GRCh38
rs780896495 Chr.20: 46178045-46178045 on Build GRCh38
rs1007971464 Chr.20: 46177890-46177890 on Build GRCh38
rs774791674 Chr.20: 46177975-46177975 on Build GRCh38
rs777782958 Chr.20: 46177919-46177919 on Build GRCh38
rs768142284 Chr.20: 46177977-46177977 on Build GRCh38
rs746538841 Chr.20: 46177976-46177976 on Build GRCh38
rs369762652 Chr.20: 46177901-46177901 on Build GRCh38
rs201893695 Chr.20: 46177937-46177937 on Build GRCh38
rs749043438 Chr.20: 46177961-46177961 on Build GRCh38
rs751728040 Chr.20: 46177944-46177944 on Build GRCh38
rs755097695 Chr.20: 46177947-46177947 on Build GRCh38
rs759021901 Chr.20: 46177935-46177935 on Build GRCh38
rs73312756 Chr.20: 46177895-46177895 on Build GRCh38
rs915961513 Chr.20: 46178002-46178002 on Build GRCh38
rs147239577 Chr.20: 46177985-46177985 on Build GRCh38

View Full Product Details