Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 19405895-19406120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSGALNACT1
Entrez Gene ID
55790
Gene Name
chondroitin sulfate N-acetylgalactosaminyltransferase 1
Gene Aliases
CSGalNAcT-1, ChGn, beta4GalNAcT
UniGene
Hs.613729
Gene Chromosome Location
Chr.8: 19404161-19757940 on Build GRCh38
Transcripts
NM_001130518.1 NM_018371.4 NR_024040.1 XM_006716358.2 XM_006716359.2 XM_006716360.2 XM_006716361.2 XM_006716362.2 XM_006716363.1 XM_006716364.3 XM_011544578.1 XM_011544579.1 XM_011544580.1 XM_011544582.1 XM_011544583.1 XM_011544584.1 XM_011544585.2 XM_017013623.1 XM_017013624.1 XM_017013625.1 XM_017013626.1 XM_017013627.1 XM_017013628.1 XM_017013629.1 XM_017013630.1 XM_017013631.1 XM_017013632.1 XM_017013633.1 XM_017013634.1 XM_017013635.1 XM_017013636.1 XM_017013637.1 XM_017013638.1 XM_017013639.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
243871 c.1414C>G p.L472V Chr.8: 19405965-19405965 on Build GRCh38
2784985 c.1481T>C p.M494T Chr.8: 19405898-19405898 on Build GRCh38
2784987 c.1396C>T p.R466W Chr.8: 19405983-19405983 on Build GRCh38
454271 c.1397G>A p.R466Q Chr.8: 19405982-19405982 on Build GRCh38
4965442 c.1341C>A p.G447G Chr.8: 19406038-19406038 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754364833 Chr.8: 19406031-19406031 on Build GRCh38
rs369585957 Chr.8: 19406013-19406013 on Build GRCh38
rs776407676 Chr.8: 19405941-19405941 on Build GRCh38
rs765103135 Chr.8: 19405954-19405954 on Build GRCh38
rs370750740 Chr.8: 19406079-19406079 on Build GRCh38
rs930411807 Chr.8: 19406014-19406014 on Build GRCh38
rs199989662 Chr.8: 19405982-19405982 on Build GRCh38
rs200462016 Chr.8: 19405970-19405970 on Build GRCh38
rs146677046 Chr.8: 19405917-19405917 on Build GRCh38
rs768320984 Chr.8: 19406040-19406040 on Build GRCh38
rs760941508 Chr.8: 19406093-19406093 on Build GRCh38
rs769525948 Chr.8: 19406058-19406058 on Build GRCh38
rs762925066 Chr.8: 19406009-19406009 on Build GRCh38
rs376968161 Chr.8: 19406016-19406016 on Build GRCh38
rs753145212 Chr.8: 19406018-19406018 on Build GRCh38
rs181623179 Chr.8: 19405966-19405966 on Build GRCh38
rs768232625 Chr.8: 19405988-19405988 on Build GRCh38
rs376979679 Chr.8: 19406075-19406075 on Build GRCh38
rs918967773 Chr.8: 19405986-19405986 on Build GRCh38
rs770746234 Chr.8: 19406050-19406050 on Build GRCh38
rs761745169 Chr.8: 19405949-19405949 on Build GRCh38
rs375826708 Chr.8: 19405965-19405965 on Build GRCh38
rs775906648 Chr.8: 19406092-19406092 on Build GRCh38
rs760302332 Chr.8: 19405937-19405937 on Build GRCh38
rs372706057 Chr.8: 19406038-19406038 on Build GRCh38
rs553914582 Chr.8: 19405918-19405918 on Build GRCh38
rs979630298 Chr.8: 19405932-19405932 on Build GRCh38
rs146627040 Chr.8: 19405931-19405931 on Build GRCh38
rs761549070 Chr.8: 19405991-19405991 on Build GRCh38
rs756700104 Chr.8: 19405977-19405977 on Build GRCh38
rs200963366 Chr.8: 19405974-19405974 on Build GRCh38
rs186245696 Chr.8: 19405993-19405993 on Build GRCh38
rs372986958 Chr.8: 19405947-19405947 on Build GRCh38
rs747767054 Chr.8: 19406055-19406055 on Build GRCh38
rs1020615160 Chr.8: 19406082-19406082 on Build GRCh38
rs1049574751 Chr.8: 19405983-19405983 on Build GRCh38
rs773046837 Chr.8: 19406069-19406069 on Build GRCh38
rs767676796 Chr.8: 19406088-19406088 on Build GRCh38
rs144295336 Chr.8: 19405940-19405940 on Build GRCh38
rs778181331 Chr.8: 19406024-19406024 on Build GRCh38
rs371393969 Chr.8: 19406023-19406023 on Build GRCh38
rs139076756 Chr.8: 19405929-19405929 on Build GRCh38
rs544410186 Chr.8: 19406034-19406034 on Build GRCh38
rs17128366 Chr.8: 19405961-19405961 on Build GRCh38
rs915608734 Chr.8: 19406042-19406042 on Build GRCh38
rs778105254 Chr.8: 19405978-19405978 on Build GRCh38
rs74336185 Chr.8: 19405934-19405934 on Build GRCh38
rs35971700 Chr.8: 19405930-19405930 on Build GRCh38
rs779606427 Chr.8: 19405984-19405984 on Build GRCh38
rs781730594 Chr.8: 19405971-19405971 on Build GRCh38
rs766427423 Chr.8: 19406012-19406012 on Build GRCh38
rs749246315 Chr.8: 19406074-19406074 on Build GRCh38
rs764437178 Chr.8: 19406094-19406094 on Build GRCh38
rs948601223 Chr.8: 19406045-19406045 on Build GRCh38
rs201889650 Chr.8: 19405960-19405960 on Build GRCh38
rs372768675 Chr.8: 19405989-19405989 on Build GRCh38
rs375237590 Chr.8: 19405995-19405995 on Build GRCh38
rs779435105 Chr.8: 19406037-19406037 on Build GRCh38
rs369428087 Chr.8: 19405979-19405979 on Build GRCh38
rs1045718996 Chr.8: 19406048-19406048 on Build GRCh38
rs536340909 Chr.8: 19406029-19406029 on Build GRCh38
rs766450784 Chr.8: 19405964-19405964 on Build GRCh38
rs148967858 Chr.8: 19406015-19406015 on Build GRCh38

View Full Product Details