Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 180632129-180632390 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XPR1
Entrez Gene ID
9213
Gene Name
xenotropic and polytropic retrovirus receptor 1
Gene Aliases
IBGC6, SYG1, X3
UniGene
Hs.227656
Gene Chromosome Location
Chr.1: 180632010-180890279 on Build GRCh38
Transcripts
NM_001135669.1 NM_004736.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
297402 c.60C>A p.I20I Chr.1: 180632261-180632261 on Build GRCh38
4967448 c.61C>T p.Q21* Chr.1: 180632262-180632262 on Build GRCh38
6824115 c.40G>A p.E14K Chr.1: 180632241-180632241 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs772195926 Chr.1: 180632319-180632319 on Build GRCh38
rs374624546 Chr.1: 180632172-180632172 on Build GRCh38
rs1038861973 Chr.1: 180632196-180632196 on Build GRCh38
rs979711921 Chr.1: 180632329-180632329 on Build GRCh38
rs902173439 Chr.1: 180632275-180632275 on Build GRCh38
rs781015863 Chr.1: 180632155-180632155 on Build GRCh38
rs114508118 Chr.1: 180632343-180632343 on Build GRCh38
rs766778205 Chr.1: 180632213-180632213 on Build GRCh38
rs940149244 Chr.1: 180632371-180632371 on Build GRCh38
rs745825410 Chr.1: 180632285-180632285 on Build GRCh38
rs537626261 Chr.1: 180632349-180632349 on Build GRCh38
rs894029451 Chr.1: 180632322-180632322 on Build GRCh38
rs774166422 Chr.1: 180632281-180632281 on Build GRCh38
rs750788810 Chr.1: 180632207-180632207 on Build GRCh38
rs996790561 Chr.1: 180632147-180632147 on Build GRCh38
rs368840063 Chr.1: 180632177-180632177 on Build GRCh38
rs984108022 Chr.1: 180632368-180632368 on Build GRCh38
rs904412158 Chr.1: 180632150-180632150 on Build GRCh38
rs371060222 Chr.1: 180632152-180632152 on Build GRCh38
rs757202895 Chr.1: 180632267-180632267 on Build GRCh38
rs752296450 Chr.1: 180632219-180632219 on Build GRCh38
rs751115742 Chr.1: 180632255-180632255 on Build GRCh38
rs548175050 Chr.1: 180632163-180632163 on Build GRCh38
rs371495767 Chr.1: 180632296-180632296 on Build GRCh38
rs369010336 Chr.1: 180632308-180632308 on Build GRCh38
rs376959497 Chr.1: 180632210-180632210 on Build GRCh38
rs758076971 Chr.1: 180632228-180632228 on Build GRCh38
rs777229584 Chr.1: 180632240-180632240 on Build GRCh38
rs926887622 Chr.1: 180632330-180632330 on Build GRCh38
rs1028262398 Chr.1: 180632149-180632149 on Build GRCh38
rs769040337 Chr.1: 180632306-180632306 on Build GRCh38
rs951442600 Chr.1: 180632354-180632354 on Build GRCh38
rs768811364 Chr.1: 180632174-180632174 on Build GRCh38
rs1012389817 Chr.1: 180632327-180632327 on Build GRCh38
rs374314804 Chr.1: 180632288-180632288 on Build GRCh38
rs781056311 Chr.1: 180632280-180632280 on Build GRCh38
rs762380166 Chr.1: 180632188-180632188 on Build GRCh38
rs983771973 Chr.1: 180632156-180632156 on Build GRCh38
rs201051930 Chr.1: 180632303-180632303 on Build GRCh38
rs767960347 Chr.1: 180632191-180632191 on Build GRCh38
rs745704635 Chr.1: 180632158-180632158 on Build GRCh38
rs570569725 Chr.1: 180632348-180632348 on Build GRCh38
rs762043339 Chr.1: 180632309-180632309 on Build GRCh38

View Full Product Details