Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 3930068-3930341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP2A3
Entrez Gene ID
489
Gene Name
ATPase sarcoplasmic/endoplasmic reticulum Ca2+ transporting 3
Gene Aliases
SERCA3
UniGene
Hs.513870
Gene Chromosome Location
Chr.17: 3923869-3964464 on Build GRCh38
Transcripts
NM_005173.3 NM_174953.2 NM_174954.2 NM_174955.2 NM_174956.2 NM_174957.2 NM_174958.2 XM_011523881.2 XM_011523882.2 XM_011523884.2 XM_011523885.1 XM_011523888.2 XM_011523889.1 XM_017024692.1 XM_017024693.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4856027 c.2728T>C p.C910R Chr.17: 3930317-3930317 on Build GRCh38
1709583 c.2705C>T p.S902F Chr.17: 3930340-3930340 on Build GRCh38
6002433 c.2710C>T p.L904F Chr.17: 3930335-3930335 on Build GRCh38
6002432 c.2710C>T p.L904F Chr.17: 3930335-3930335 on Build GRCh38
1660689 c.2707G>A p.V903M Chr.17: 3930338-3930338 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146043310 Chr.17: 3930312-3930312 on Build GRCh38
rs71381537 Chr.17: 3930104-3930105 on Build GRCh38
rs71381534 Chr.17: 3930097-3930098 on Build GRCh38
rs866361194 Chr.17: 3930311-3930311 on Build GRCh38
rs1045865552 Chr.17: 3930141-3930141 on Build GRCh38
rs745470009 Chr.17: 3930260-3930260 on Build GRCh38
rs59380955 Chr.17: 3930104-3930104 on Build GRCh38
rs376994562 Chr.17: 3930277-3930277 on Build GRCh38
rs71381539 Chr.17: 3930108-3930109 on Build GRCh38
rs929256460 Chr.17: 3930230-3930230 on Build GRCh38
rs59833705 Chr.17: 3930235-3930235 on Build GRCh38
rs542916351 Chr.17: 3930169-3930169 on Build GRCh38
rs752379112 Chr.17: 3930308-3930308 on Build GRCh38
rs67622655 Chr.17: 3930206-3930297 on Build GRCh38
rs71381540 Chr.17: 3930113-3930114 on Build GRCh38
rs775648328 Chr.17: 3930262-3930262 on Build GRCh38
rs777570900 Chr.17: 3930321-3930321 on Build GRCh38
rs71381538 Chr.17: 3930107-3930108 on Build GRCh38
rs375596476 Chr.17: 3930106-3930106 on Build GRCh38
rs1004212148 Chr.17: 3930198-3930198 on Build GRCh38
rs773967641 Chr.17: 3930257-3930257 on Build GRCh38
rs2240805 Chr.17: 3930177-3930177 on Build GRCh38
rs770498756 Chr.17: 3930254-3930254 on Build GRCh38
rs980340908 Chr.17: 3930114-3930114 on Build GRCh38
rs928605139 Chr.17: 3930124-3930124 on Build GRCh38
rs57591991 Chr.17: 3930206-3930206 on Build GRCh38
rs765457449 Chr.17: 3930283-3930283 on Build GRCh38
rs56164329 Chr.17: 3930207-3930208 on Build GRCh38
rs8074234 Chr.17: 3930193-3930193 on Build GRCh38
rs977806566 Chr.17: 3930313-3930313 on Build GRCh38
rs771741280 Chr.17: 3930261-3930261 on Build GRCh38
rs565369204 Chr.17: 3930197-3930197 on Build GRCh38
rs766715327 Chr.17: 3930299-3930299 on Build GRCh38
rs779392080 Chr.17: 3930130-3930130 on Build GRCh38
rs71381535 Chr.17: 3930098-3930099 on Build GRCh38
rs548979977 Chr.17: 3930275-3930275 on Build GRCh38
rs71381536 Chr.17: 3930100-3930101 on Build GRCh38
rs761112190 Chr.17: 3930268-3930268 on Build GRCh38
rs12453032 Chr.17: 3930269-3930269 on Build GRCh38
rs139066738 Chr.17: 3930246-3930246 on Build GRCh38
rs71381533 Chr.17: 3930096-3930097 on Build GRCh38
rs763228999 Chr.17: 3930289-3930289 on Build GRCh38
rs374825886 Chr.17: 3930291-3930291 on Build GRCh38
rs1019425639 Chr.17: 3930290-3930290 on Build GRCh38
rs916146890 Chr.17: 3930126-3930126 on Build GRCh38
rs1056504829 Chr.17: 3930192-3930192 on Build GRCh38
rs186613169 Chr.17: 3930182-3930182 on Build GRCh38
rs370125024 Chr.17: 3930287-3930287 on Build GRCh38
rs966537166 Chr.17: 3930298-3930298 on Build GRCh38
rs765405100 Chr.17: 3930176-3930181 on Build GRCh38
rs1037096903 Chr.17: 3930225-3930225 on Build GRCh38
rs998195246 Chr.17: 3930300-3930300 on Build GRCh38
rs748739168 Chr.17: 3930252-3930252 on Build GRCh38
rs1055227320 Chr.17: 3930105-3930105 on Build GRCh38

View Full Product Details