Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 121113615-121113840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTRL
Entrez Gene ID
11064
Gene Name
centriolin
Gene Aliases
CEP1, CEP110, FAN, bA165P4.1
UniGene
Hs.653263
Gene Chromosome Location
Chr.9: 121074778-121177610 on Build GRCh38
Transcripts
NM_007018.4 XM_005251679.3 XM_006716940.2 XM_006716944.3 XM_011518166.1 XM_011518167.1 XM_011518168.1 XM_011518169.2 XM_011518170.1 XM_011518172.2 XM_017014223.1 XM_017014224.1 XM_017014225.1 XM_017014227.1 XM_017014228.1 XM_017014229.1 XM_017014230.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6039252 c.1325A>G p.K442R Chr.9: 121113704-121113704 on Build GRCh38
6632005 c.1329A>C p.E443D Chr.9: 121113708-121113708 on Build GRCh38
3322780 c.1311G>A p.T437T Chr.9: 121113690-121113690 on Build GRCh38
1459732 c.1328delA p.I446fs*1 Chr.9: 121113707-121113707 on Build GRCh38
1624494 c.1256A>G p.D419G Chr.9: 121113635-121113635 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs983093653 Chr.9: 121113721-121113721 on Build GRCh38
rs758278344 Chr.9: 121113658-121113658 on Build GRCh38
rs765875810 Chr.9: 121113660-121113660 on Build GRCh38
rs951418960 Chr.9: 121113720-121113720 on Build GRCh38
rs1043166437 Chr.9: 121113771-121113771 on Build GRCh38
rs781258094 Chr.9: 121113728-121113729 on Build GRCh38
rs776672961 Chr.9: 121113703-121113703 on Build GRCh38
rs754643567 Chr.9: 121113745-121113745 on Build GRCh38
rs916672062 Chr.9: 121113751-121113751 on Build GRCh38
rs746154108 Chr.9: 121113682-121113682 on Build GRCh38
rs780880088 Chr.9: 121113670-121113670 on Build GRCh38
rs757230210 Chr.9: 121113715-121113716 on Build GRCh38
rs397840585 Chr.9: 121113760-121113760 on Build GRCh38
rs755692655 Chr.9: 121113759-121113759 on Build GRCh38
rs761941553 Chr.9: 121113708-121113708 on Build GRCh38
rs770310579 Chr.9: 121113751-121113752 on Build GRCh38
rs547352269 Chr.9: 121113689-121113689 on Build GRCh38
rs758616858 Chr.9: 121113703-121113706 on Build GRCh38
rs780439558 Chr.9: 121113776-121113776 on Build GRCh38
rs138886404 Chr.9: 121113668-121113668 on Build GRCh38
rs879554252 Chr.9: 121113790-121113790 on Build GRCh38
rs751131421 Chr.9: 121113742-121113742 on Build GRCh38
rs563715883 Chr.9: 121113779-121113779 on Build GRCh38
rs751234775 Chr.9: 121113666-121113666 on Build GRCh38
rs77083332 Chr.9: 121113643-121113643 on Build GRCh38
rs3831035 Chr.9: 121113752-121113752 on Build GRCh38
rs1035582420 Chr.9: 121113707-121113710 on Build GRCh38
rs960225406 Chr.9: 121113748-121113748 on Build GRCh38
rs763230429 Chr.9: 121113657-121113657 on Build GRCh38
rs189082329 Chr.9: 121113691-121113691 on Build GRCh38
rs757815527 Chr.9: 121113678-121113678 on Build GRCh38
rs773202160 Chr.9: 121113719-121113719 on Build GRCh38
rs370735215 Chr.9: 121113703-121113705 on Build GRCh38
rs767325490 Chr.9: 121113747-121113747 on Build GRCh38
rs777925006 Chr.9: 121113706-121113707 on Build GRCh38
rs145070862 Chr.9: 121113654-121113654 on Build GRCh38
rs750962721 Chr.9: 121113761-121113761 on Build GRCh38
rs762593932 Chr.9: 121113725-121113725 on Build GRCh38
rs747099268 Chr.9: 121113707-121113707 on Build GRCh38
rs765211287 Chr.9: 121113647-121113647 on Build GRCh38
rs142008519 Chr.9: 121113671-121113671 on Build GRCh38
rs557327937 Chr.9: 121113690-121113690 on Build GRCh38
rs766218059 Chr.9: 121113735-121113735 on Build GRCh38
rs370202924 Chr.9: 121113688-121113688 on Build GRCh38
rs546133951 Chr.9: 121113752-121113752 on Build GRCh38
rs765152044 Chr.9: 121113713-121113713 on Build GRCh38
rs1026993673 Chr.9: 121113717-121113717 on Build GRCh38
rs758654056 Chr.9: 121113770-121113770 on Build GRCh38

View Full Product Details