Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 101164620-101164883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VGF
Entrez Gene ID
7425
Gene Name
VGF nerve growth factor inducible
Gene Aliases
SCG7, SgVII
UniGene
Hs.587325
Gene Chromosome Location
Chr.7: 101162509-101169956 on Build GRCh38
Transcripts
NM_003378.3 XM_005250561.4 XM_011516549.2 XM_017012579.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6350157 c.26C>A p.S9Y Chr.7: 101164818-101164818 on Build GRCh38
1083386 c.51C>A p.I17I Chr.7: 101164793-101164793 on Build GRCh38
3358070 c.98C>T p.P33L Chr.7: 101164746-101164746 on Build GRCh38
6515382 c.8C>T p.A3V Chr.7: 101164836-101164836 on Build GRCh38
4595973 c.198G>T p.E66D Chr.7: 101164646-101164646 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775796913 Chr.7: 101164739-101164739 on Build GRCh38
rs376363797 Chr.7: 101164782-101164782 on Build GRCh38
rs764198367 Chr.7: 101164643-101164643 on Build GRCh38
rs755864356 Chr.7: 101164716-101164716 on Build GRCh38
rs780164922 Chr.7: 101164688-101164688 on Build GRCh38
rs139767719 Chr.7: 101164828-101164828 on Build GRCh38
rs376279036 Chr.7: 101164685-101164685 on Build GRCh38
rs767562242 Chr.7: 101164795-101164795 on Build GRCh38
rs777510124 Chr.7: 101164824-101164824 on Build GRCh38
rs147256753 Chr.7: 101164759-101164759 on Build GRCh38
rs932310732 Chr.7: 101164846-101164846 on Build GRCh38
rs772383358 Chr.7: 101164736-101164736 on Build GRCh38
rs765508638 Chr.7: 101164751-101164751 on Build GRCh38
rs747559487 Chr.7: 101164741-101164741 on Build GRCh38
rs755028529 Chr.7: 101164774-101164774 on Build GRCh38
rs761731782 Chr.7: 101164748-101164748 on Build GRCh38
rs368851042 Chr.7: 101164718-101164718 on Build GRCh38
rs372545984 Chr.7: 101164746-101164746 on Build GRCh38
rs908365438 Chr.7: 101164771-101164771 on Build GRCh38
rs755523412 Chr.7: 101164822-101164822 on Build GRCh38
rs776837131 Chr.7: 101164747-101164747 on Build GRCh38
rs1002621254 Chr.7: 101164726-101164726 on Build GRCh38
rs756877078 Chr.7: 101164836-101164836 on Build GRCh38
rs1035089684 Chr.7: 101164741-101164743 on Build GRCh38
rs765369283 Chr.7: 101164651-101164651 on Build GRCh38
rs762149695 Chr.7: 101164650-101164650 on Build GRCh38
rs750234248 Chr.7: 101164675-101164675 on Build GRCh38
rs1020817642 Chr.7: 101164768-101164768 on Build GRCh38
rs752259121 Chr.7: 101164813-101164813 on Build GRCh38
rs989692287 Chr.7: 101164762-101164762 on Build GRCh38
rs747598672 Chr.7: 101164711-101164711 on Build GRCh38
rs747368903 Chr.7: 101164858-101164858 on Build GRCh38
rs751505743 Chr.7: 101164696-101164696 on Build GRCh38
rs753892644 Chr.7: 101164645-101164645 on Build GRCh38
rs773260623 Chr.7: 101164754-101164754 on Build GRCh38
rs545909485 Chr.7: 101164773-101164773 on Build GRCh38
rs746260665 Chr.7: 101164723-101164723 on Build GRCh38
rs780902669 Chr.7: 101164709-101164709 on Build GRCh38
rs1024575484 Chr.7: 101164641-101164641 on Build GRCh38
rs774311991 Chr.7: 101164857-101164857 on Build GRCh38
rs754938691 Chr.7: 101164697-101164697 on Build GRCh38

View Full Product Details