Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 20384616-20384861 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDILT
Entrez Gene ID
204474
Gene Name
protein disulfide isomerase like, testis expressed
Gene Aliases
PDIA7
UniGene
Hs.376025
Gene Chromosome Location
Chr.16: 20359170-20404742 on Build GRCh38
Transcripts
NM_174924.1 XM_011545764.1 XM_011545765.1 XM_011545766.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6555493 c.294T>A p.F98L Chr.16: 20384760-20384760 on Build GRCh38
6002179 c.266T>C p.M89T Chr.16: 20384788-20384788 on Build GRCh38
968045 c.227G>A p.R76K Chr.16: 20384827-20384827 on Build GRCh38
6586095 c.311C>T p.T104I Chr.16: 20384743-20384743 on Build GRCh38
1172311 c.389C>A p.S130* Chr.16: 20384665-20384665 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs930303262 Chr.16: 20384644-20384644 on Build GRCh38
rs773672538 Chr.16: 20384684-20384684 on Build GRCh38
rs779933955 Chr.16: 20384780-20384780 on Build GRCh38
rs907016907 Chr.16: 20384706-20384706 on Build GRCh38
rs534001534 Chr.16: 20384794-20384794 on Build GRCh38
rs757076614 Chr.16: 20384710-20384710 on Build GRCh38
rs763323272 Chr.16: 20384667-20384667 on Build GRCh38
rs760000909 Chr.16: 20384772-20384772 on Build GRCh38
rs377332564 Chr.16: 20384658-20384658 on Build GRCh38
rs778925498 Chr.16: 20384717-20384717 on Build GRCh38
rs539440383 Chr.16: 20384719-20384719 on Build GRCh38
rs755877564 Chr.16: 20384697-20384697 on Build GRCh38
rs934794878 Chr.16: 20384707-20384707 on Build GRCh38
rs566608055 Chr.16: 20384766-20384766 on Build GRCh38
rs767200436 Chr.16: 20384689-20384689 on Build GRCh38
rs773620193 Chr.16: 20384660-20384660 on Build GRCh38
rs146719463 Chr.16: 20384837-20384837 on Build GRCh38
rs375479299 Chr.16: 20384765-20384765 on Build GRCh38
rs150624587 Chr.16: 20384700-20384700 on Build GRCh38
rs753509112 Chr.16: 20384670-20384670 on Build GRCh38
rs893178850 Chr.16: 20384757-20384757 on Build GRCh38
rs1024003682 Chr.16: 20384664-20384664 on Build GRCh38
rs773495613 Chr.16: 20384738-20384738 on Build GRCh38
rs139688634 Chr.16: 20384735-20384735 on Build GRCh38
rs1019771750 Chr.16: 20384803-20384803 on Build GRCh38
rs755084345 Chr.16: 20384726-20384726 on Build GRCh38
rs866135832 Chr.16: 20384720-20384720 on Build GRCh38
rs775162102 Chr.16: 20384789-20384789 on Build GRCh38
rs750346707 Chr.16: 20384828-20384828 on Build GRCh38
rs149811393 Chr.16: 20384736-20384736 on Build GRCh38
rs766910703 Chr.16: 20384672-20384672 on Build GRCh38
rs555849933 Chr.16: 20384817-20384817 on Build GRCh38
rs141023477 Chr.16: 20384806-20384806 on Build GRCh38
rs781291960 Chr.16: 20384731-20384731 on Build GRCh38
rs142965458 Chr.16: 20384687-20384687 on Build GRCh38
rs140161685 Chr.16: 20384656-20384656 on Build GRCh38
rs890052139 Chr.16: 20384792-20384792 on Build GRCh38
rs143343359 Chr.16: 20384818-20384818 on Build GRCh38
rs746922029 Chr.16: 20384749-20384767 on Build GRCh38
rs749672868 Chr.16: 20384753-20384753 on Build GRCh38
rs1020428645 Chr.16: 20384787-20384787 on Build GRCh38
rs776822199 Chr.16: 20384653-20384653 on Build GRCh38
rs779987113 Chr.16: 20384678-20384679 on Build GRCh38
rs752364490 Chr.16: 20384695-20384695 on Build GRCh38
rs201976699 Chr.16: 20384698-20384698 on Build GRCh38

View Full Product Details