Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 39898367-39898624 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCP4
Entrez Gene ID
5121
Gene Name
Purkinje cell protein 4
Gene Aliases
PEP-19
UniGene
Hs.80296
Gene Chromosome Location
Chr.21: 39867421-39929397 on Build GRCh38
Transcripts
NM_006198.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3693790 c.11G>A p.R4Q Chr.21: 39898477-39898477 on Build GRCh38
4101638 c.25G>A p.A9T Chr.21: 39898491-39898491 on Build GRCh38
1030783 c.40G>T p.D14Y Chr.21: 39898506-39898506 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753978459 Chr.21: 39898555-39898555 on Build GRCh38
rs761172054 Chr.21: 39898537-39898537 on Build GRCh38
rs866112942 Chr.21: 39898527-39898527 on Build GRCh38
rs774719967 Chr.21: 39898540-39898540 on Build GRCh38
rs776072472 Chr.21: 39898407-39898407 on Build GRCh38
rs961443237 Chr.21: 39898458-39898458 on Build GRCh38
rs755287934 Chr.21: 39898432-39898432 on Build GRCh38
rs376912296 Chr.21: 39898572-39898572 on Build GRCh38
rs927538965 Chr.21: 39898506-39898506 on Build GRCh38
rs200487465 Chr.21: 39898529-39898529 on Build GRCh38
rs746686736 Chr.21: 39898513-39898513 on Build GRCh38
rs769017835 Chr.21: 39898588-39898588 on Build GRCh38
rs759989792 Chr.21: 39898544-39898544 on Build GRCh38
rs756307513 Chr.21: 39898455-39898455 on Build GRCh38
rs749338940 Chr.21: 39898473-39898473 on Build GRCh38
rs768259290 Chr.21: 39898514-39898514 on Build GRCh38
rs778974674 Chr.21: 39898563-39898563 on Build GRCh38
rs776125601 Chr.21: 39898528-39898528 on Build GRCh38
rs771240699 Chr.21: 39898449-39898450 on Build GRCh38
rs773699802 Chr.21: 39898407-39898407 on Build GRCh38
rs753059047 Chr.21: 39898550-39898550 on Build GRCh38
rs766732498 Chr.21: 39898428-39898428 on Build GRCh38
rs747184398 Chr.21: 39898580-39898580 on Build GRCh38
rs777000487 Chr.21: 39898461-39898461 on Build GRCh38
rs757269012 Chr.21: 39898560-39898560 on Build GRCh38
rs144011084 Chr.21: 39898476-39898476 on Build GRCh38
rs767747233 Chr.21: 39898548-39898548 on Build GRCh38
rs764342171 Chr.21: 39898552-39898552 on Build GRCh38
rs771760041 Chr.21: 39898483-39898483 on Build GRCh38
rs377232216 Chr.21: 39898439-39898439 on Build GRCh38
rs779857117 Chr.21: 39898576-39898576 on Build GRCh38
rs771529727 Chr.21: 39898538-39898538 on Build GRCh38
rs939101379 Chr.21: 39898579-39898579 on Build GRCh38
rs761006617 Chr.21: 39898551-39898551 on Build GRCh38
rs933669957 Chr.21: 39898599-39898599 on Build GRCh38
rs751969308 Chr.21: 39898429-39898429 on Build GRCh38
rs957426927 Chr.21: 39898420-39898420 on Build GRCh38
rs946904367 Chr.21: 39898539-39898539 on Build GRCh38
rs940841192 Chr.21: 39898592-39898592 on Build GRCh38
rs750306398 Chr.21: 39898570-39898570 on Build GRCh38
rs185480998 Chr.21: 39898427-39898427 on Build GRCh38
rs896884864 Chr.21: 39898597-39898597 on Build GRCh38
rs986124920 Chr.21: 39898515-39898515 on Build GRCh38
rs747551110 Chr.21: 39898441-39898443 on Build GRCh38
rs369716340 Chr.21: 39898441-39898441 on Build GRCh38
rs745733374 Chr.21: 39898477-39898477 on Build GRCh38
rs569706286 Chr.21: 39898603-39898603 on Build GRCh38
rs778018654 Chr.21: 39898461-39898461 on Build GRCh38
rs2075719 Chr.21: 39898596-39898596 on Build GRCh38
rs374105114 Chr.21: 39898498-39898498 on Build GRCh38
rs551897534 Chr.21: 39898422-39898422 on Build GRCh38

View Full Product Details