Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 74700202-74700461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2I
Entrez Gene ID
2969
Gene Name
general transcription factor IIi
Gene Aliases
BAP135, BTKAP1, DIWS, GTFII-I, IB291, SPIN, TFII-I, WBS, WBSCR6
UniGene
Hs.647041
Gene Chromosome Location
Chr.7: 74657665-74760692 on Build GRCh38
Transcripts
NM_001163636.2 NM_001280800.1 NM_001518.4 NM_032999.3 NM_033000.3 NM_033001.3 NR_110044.1

Gene Symbol
LOC101926943
Entrez Gene ID
101926943
Gene Name
uncharacterized LOC101926943
Gene Aliases
-
UniGene
Hs.654772
Gene Chromosome Location
Chr.7: 74688937-74728918 on Build GRCh38
Transcripts
NM_001163636.2 NM_001280800.1 NM_001518.4 NM_032999.3 NM_033000.3 NM_033001.3 NR_110044.1

SNP Detail

SNP ID Chromosome Location
rs782514449 Chr.7: 74700299-74700299 on Build GRCh38
rs782485177 Chr.7: 74700368-74700368 on Build GRCh38
rs782793898 Chr.7: 74700346-74700346 on Build GRCh38
rs990392390 Chr.7: 74700390-74700390 on Build GRCh38
rs1038690408 Chr.7: 74700229-74700229 on Build GRCh38
rs1064765 Chr.7: 74700300-74700300 on Build GRCh38
rs374937893 Chr.7: 74700237-74700237 on Build GRCh38
rs782450205 Chr.7: 74700308-74700308 on Build GRCh38
rs782745853 Chr.7: 74700278-74700278 on Build GRCh38
rs1057896 Chr.7: 74700393-74700393 on Build GRCh38
rs1135648 Chr.7: 74700406-74700406 on Build GRCh38
rs781943013 Chr.7: 74700269-74700269 on Build GRCh38
rs781823900 Chr.7: 74700284-74700284 on Build GRCh38
rs782799288 Chr.7: 74700257-74700257 on Build GRCh38
rs782228824 Chr.7: 74700309-74700309 on Build GRCh38
rs782411031 Chr.7: 74700315-74700315 on Build GRCh38
rs782080046 Chr.7: 74700344-74700344 on Build GRCh38
rs782481420 Chr.7: 74700310-74700310 on Build GRCh38
rs781795612 Chr.7: 74700265-74700265 on Build GRCh38
rs782730755 Chr.7: 74700380-74700380 on Build GRCh38
rs782015737 Chr.7: 74700325-74700325 on Build GRCh38
rs148927442 Chr.7: 74700359-74700359 on Build GRCh38
rs781985101 Chr.7: 74700347-74700347 on Build GRCh38
rs782204993 Chr.7: 74700334-74700334 on Build GRCh38
rs145729076 Chr.7: 74700301-74700301 on Build GRCh38
rs587656081 Chr.7: 74700360-74700360 on Build GRCh38
rs781788087 Chr.7: 74700362-74700362 on Build GRCh38
rs1049009087 Chr.7: 74700297-74700297 on Build GRCh38
rs781983673 Chr.7: 74700248-74700248 on Build GRCh38
rs782057203 Chr.7: 74700270-74700270 on Build GRCh38
rs202074864 Chr.7: 74700235-74700235 on Build GRCh38

View Full Product Details