Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 183505183-183505442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ING2
Entrez Gene ID
3622
Gene Name
inhibitor of growth family member 2
Gene Aliases
ING1L, p33ING2
UniGene
Hs.107153
Gene Chromosome Location
Chr.4: 183505050-183512428 on Build GRCh38
Transcripts
NM_001564.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5639504 c.1A>G p.M1V Chr.4: 183505196-183505196 on Build GRCh38
6685843 c.49A>G p.T17A Chr.4: 183505244-183505244 on Build GRCh38
1738957 c.120C>G p.H40Q Chr.4: 183505315-183505315 on Build GRCh38
3380880 c.54G>A p.G18G Chr.4: 183505249-183505249 on Build GRCh38
1695293 c.116_117CC>TT p.P39L Chr.4: 183505311-183505312 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs6552675 Chr.4: 183505373-183505373 on Build GRCh38
rs750041352 Chr.4: 183505370-183505370 on Build GRCh38
rs774813457 Chr.4: 183505273-183505273 on Build GRCh38
rs571719648 Chr.4: 183505224-183505224 on Build GRCh38
rs771210792 Chr.4: 183505383-183505383 on Build GRCh38
rs763371219 Chr.4: 183505208-183505208 on Build GRCh38
rs11545693 Chr.4: 183505312-183505312 on Build GRCh38
rs767874568 Chr.4: 183505221-183505221 on Build GRCh38
rs758125498 Chr.4: 183505411-183505412 on Build GRCh38
rs765122329 Chr.4: 183505270-183505272 on Build GRCh38
rs760931507 Chr.4: 183505288-183505288 on Build GRCh38
rs912670082 Chr.4: 183505404-183505404 on Build GRCh38
rs763685216 Chr.4: 183505306-183505306 on Build GRCh38
rs1013704160 Chr.4: 183505338-183505338 on Build GRCh38
rs761492555 Chr.4: 183505226-183505228 on Build GRCh38
rs8872 Chr.4: 183505234-183505234 on Build GRCh38
rs766747609 Chr.4: 183505215-183505215 on Build GRCh38
rs780032686 Chr.4: 183505357-183505357 on Build GRCh38
rs763983178 Chr.4: 183505204-183505206 on Build GRCh38
rs759773632 Chr.4: 183505279-183505279 on Build GRCh38
rs778379287 Chr.4: 183505313-183505313 on Build GRCh38
rs757060299 Chr.4: 183505231-183505231 on Build GRCh38
rs989222941 Chr.4: 183505419-183505419 on Build GRCh38
rs749324061 Chr.4: 183505382-183505382 on Build GRCh38
rs753537081 Chr.4: 183505228-183505228 on Build GRCh38
rs766142641 Chr.4: 183505419-183505420 on Build GRCh38
rs752052254 Chr.4: 183505216-183505216 on Build GRCh38
rs779797468 Chr.4: 183505246-183505246 on Build GRCh38
rs755631655 Chr.4: 183505222-183505222 on Build GRCh38
rs953901627 Chr.4: 183505388-183505388 on Build GRCh38
rs567832274 Chr.4: 183505285-183505285 on Build GRCh38
rs770015934 Chr.4: 183505202-183505202 on Build GRCh38
rs772322503 Chr.4: 183505407-183505407 on Build GRCh38
rs753596147 Chr.4: 183505309-183505309 on Build GRCh38
rs1015220468 Chr.4: 183505372-183505372 on Build GRCh38
rs777797153 Chr.4: 183505376-183505376 on Build GRCh38
rs750244453 Chr.4: 183505315-183505315 on Build GRCh38
rs776572860 Chr.4: 183505203-183505204 on Build GRCh38
rs550859084 Chr.4: 183505262-183505262 on Build GRCh38
rs772067392 Chr.4: 183505245-183505245 on Build GRCh38
rs906003877 Chr.4: 183505250-183505250 on Build GRCh38
rs985245885 Chr.4: 183505400-183505400 on Build GRCh38
rs919775808 Chr.4: 183505387-183505387 on Build GRCh38
rs1053115496 Chr.4: 183505211-183505211 on Build GRCh38
rs763137621 Chr.4: 183505265-183505265 on Build GRCh38
rs778747947 Chr.4: 183505232-183505232 on Build GRCh38
rs746252654 Chr.4: 183505391-183505391 on Build GRCh38
rs769856799 Chr.4: 183505253-183505253 on Build GRCh38
rs753192153 Chr.4: 183505297-183505297 on Build GRCh38
rs758061494 Chr.4: 183505331-183505331 on Build GRCh38
rs138103277 Chr.4: 183505207-183505207 on Build GRCh38
rs372587337 Chr.4: 183505272-183505272 on Build GRCh38
rs746931867 Chr.4: 183505364-183505364 on Build GRCh38

View Full Product Details