Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 156950428-156950679 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOM1
Entrez Gene ID
64434
Gene Name
nucleolar protein with MIF4G domain 1
Gene Aliases
C7orf3, PPP1R113, SGD1
UniGene
Hs.15825
Gene Chromosome Location
Chr.7: 156949723-156973182 on Build GRCh38
Transcripts
NM_138400.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5555220 c.913G>A p.G305R Chr.7: 156950650-156950650 on Build GRCh38
366865 c.741C>T p.P247P Chr.7: 156950478-156950478 on Build GRCh38
745717 c.790G>T p.E264* Chr.7: 156950527-156950527 on Build GRCh38
4919358 c.828A>T p.A276A Chr.7: 156950565-156950565 on Build GRCh38
1449652 c.873delG p.E293fs*34 Chr.7: 156950610-156950610 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1024588089 Chr.7: 156950623-156950623 on Build GRCh38
rs62637704 Chr.7: 156950508-156950508 on Build GRCh38
rs745321468 Chr.7: 156950602-156950602 on Build GRCh38
rs370964359 Chr.7: 156950494-156950494 on Build GRCh38
rs749409847 Chr.7: 156950480-156950480 on Build GRCh38
rs116778582 Chr.7: 156950452-156950452 on Build GRCh38
rs769205294 Chr.7: 156950608-156950608 on Build GRCh38
rs746838716 Chr.7: 156950590-156950590 on Build GRCh38
rs575707726 Chr.7: 156950473-156950473 on Build GRCh38
rs964625714 Chr.7: 156950644-156950644 on Build GRCh38
rs926986345 Chr.7: 156950490-156950490 on Build GRCh38
rs756673104 Chr.7: 156950475-156950475 on Build GRCh38
rs772942362 Chr.7: 156950624-156950624 on Build GRCh38
rs146476387 Chr.7: 156950572-156950572 on Build GRCh38
rs750855725 Chr.7: 156950530-156950530 on Build GRCh38
rs748330341 Chr.7: 156950580-156950580 on Build GRCh38
rs776729133 Chr.7: 156950598-156950598 on Build GRCh38
rs374444986 Chr.7: 156950560-156950560 on Build GRCh38
rs746926718 Chr.7: 156950499-156950499 on Build GRCh38
rs773223957 Chr.7: 156950497-156950497 on Build GRCh38
rs887790903 Chr.7: 156950573-156950573 on Build GRCh38
rs540622203 Chr.7: 156950515-156950517 on Build GRCh38
rs766920341 Chr.7: 156950541-156950541 on Build GRCh38
rs768902140 Chr.7: 156950481-156950481 on Build GRCh38
rs61740418 Chr.7: 156950463-156950463 on Build GRCh38
rs775014298 Chr.7: 156950514-156950514 on Build GRCh38
rs775126294 Chr.7: 156950619-156950619 on Build GRCh38
rs994091053 Chr.7: 156950621-156950621 on Build GRCh38
rs755044233 Chr.7: 156950569-156950569 on Build GRCh38
rs764907170 Chr.7: 156950509-156950509 on Build GRCh38
rs773544472 Chr.7: 156950649-156950649 on Build GRCh38
rs767858669 Chr.7: 156950521-156950521 on Build GRCh38
rs151324588 Chr.7: 156950479-156950479 on Build GRCh38
rs756584985 Chr.7: 156950536-156950536 on Build GRCh38
rs770966157 Chr.7: 156950594-156950594 on Build GRCh38
rs913443714 Chr.7: 156950500-156950500 on Build GRCh38
rs779388601 Chr.7: 156950609-156950610 on Build GRCh38
rs758702897 Chr.7: 156950587-156950587 on Build GRCh38
rs776246212 Chr.7: 156950506-156950506 on Build GRCh38
rs762510432 Chr.7: 156950622-156950622 on Build GRCh38
rs1016863571 Chr.7: 156950610-156950610 on Build GRCh38
rs747934881 Chr.7: 156950495-156950495 on Build GRCh38
rs961629571 Chr.7: 156950612-156950612 on Build GRCh38
rs767462765 Chr.7: 156950514-156950515 on Build GRCh38
rs145298188 Chr.7: 156950533-156950533 on Build GRCh38
rs61747373 Chr.7: 156950570-156950570 on Build GRCh38
rs763778766 Chr.7: 156950632-156950632 on Build GRCh38
rs369954829 Chr.7: 156950589-156950589 on Build GRCh38
rs762182460 Chr.7: 156950515-156950515 on Build GRCh38
rs748784760 Chr.7: 156950555-156950555 on Build GRCh38
rs927019265 Chr.7: 156950650-156950650 on Build GRCh38
rs139803899 Chr.7: 156950449-156950449 on Build GRCh38
rs781617003 Chr.7: 156950464-156950464 on Build GRCh38
rs61736859 Chr.7: 156950534-156950534 on Build GRCh38
rs753453285 Chr.7: 156950448-156950448 on Build GRCh38

View Full Product Details