Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 31140151-31140386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLA2G3
Entrez Gene ID
50487
Gene Name
phospholipase A2 group III
Gene Aliases
GIII-SPLA2, SPLA2III, sPLA2-III
UniGene
Hs.149623
Gene Chromosome Location
Chr.22: 31134807-31140607 on Build GRCh38
Transcripts
NM_015715.4 XM_011530204.1 XM_011530205.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5894722 c.65C>T p.P22L Chr.22: 31140290-31140290 on Build GRCh38
1220842 c.47T>C p.V16A Chr.22: 31140308-31140308 on Build GRCh38
7002940 c.179C>A p.A60D Chr.22: 31140176-31140176 on Build GRCh38
4954976 c.35G>A p.G12D Chr.22: 31140320-31140320 on Build GRCh38
2937997 c.106G>A p.A36T Chr.22: 31140249-31140249 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757252812 Chr.22: 31140218-31140218 on Build GRCh38
rs771503708 Chr.22: 31140277-31140277 on Build GRCh38
rs745515916 Chr.22: 31140169-31140169 on Build GRCh38
rs201934732 Chr.22: 31140173-31140173 on Build GRCh38
rs772449841 Chr.22: 31140252-31140252 on Build GRCh38
rs200842920 Chr.22: 31140366-31140366 on Build GRCh38
rs777279506 Chr.22: 31140281-31140281 on Build GRCh38
rs147658733 Chr.22: 31140208-31140208 on Build GRCh38
rs371471371 Chr.22: 31140182-31140182 on Build GRCh38
rs750361564 Chr.22: 31140325-31140325 on Build GRCh38
rs548774719 Chr.22: 31140361-31140361 on Build GRCh38
rs540358884 Chr.22: 31140300-31140300 on Build GRCh38
rs569648138 Chr.22: 31140365-31140365 on Build GRCh38
rs200040239 Chr.22: 31140342-31140342 on Build GRCh38
rs138057115 Chr.22: 31140179-31140179 on Build GRCh38
rs142353712 Chr.22: 31140246-31140246 on Build GRCh38
rs188950582 Chr.22: 31140174-31140174 on Build GRCh38
rs2232175 Chr.22: 31140247-31140247 on Build GRCh38
rs781690498 Chr.22: 31140359-31140359 on Build GRCh38
rs745327229 Chr.22: 31140231-31140231 on Build GRCh38
rs559972453 Chr.22: 31140310-31140310 on Build GRCh38
rs775762409 Chr.22: 31140291-31140291 on Build GRCh38
rs759545229 Chr.22: 31140192-31140192 on Build GRCh38
rs780667204 Chr.22: 31140222-31140222 on Build GRCh38
rs767378177 Chr.22: 31140311-31140311 on Build GRCh38
rs752896259 Chr.22: 31140197-31140197 on Build GRCh38
rs753520211 Chr.22: 31140331-31140331 on Build GRCh38
rs771452463 Chr.22: 31140353-31140353 on Build GRCh38
rs765256970 Chr.22: 31140290-31140290 on Build GRCh38
rs368637435 Chr.22: 31140298-31140298 on Build GRCh38
rs933399477 Chr.22: 31140299-31140299 on Build GRCh38
rs373821255 Chr.22: 31140202-31140202 on Build GRCh38
rs140320450 Chr.22: 31140334-31140334 on Build GRCh38
rs201571423 Chr.22: 31140262-31140262 on Build GRCh38
rs896133812 Chr.22: 31140309-31140309 on Build GRCh38
rs759618441 Chr.22: 31140286-31140286 on Build GRCh38
rs776905378 Chr.22: 31140183-31140183 on Build GRCh38
rs773780010 Chr.22: 31140259-31140259 on Build GRCh38
rs528919853 Chr.22: 31140328-31140328 on Build GRCh38
rs747309168 Chr.22: 31140348-31140348 on Build GRCh38
rs763126255 Chr.22: 31140296-31140296 on Build GRCh38
rs556762951 Chr.22: 31140181-31140181 on Build GRCh38
rs545672880 Chr.22: 31140212-31140212 on Build GRCh38
rs765430625 Chr.22: 31140195-31140195 on Build GRCh38

View Full Product Details