Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 111371524-111371795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0368
Entrez Gene ID
23392
Gene Name
KIAA0368
Gene Aliases
ECM29
UniGene
Hs.368255
Gene Chromosome Location
Chr.9: 111360693-111484745 on Build GRCh38
Transcripts
NM_001080398.1 XM_005251850.3 XM_005251851.4 XM_005251852.4 XM_005251853.3 XM_005251854.3 XM_011518422.2 XM_017014557.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1255559 c.5169G>A p.M1723I Chr.9: 111371723-111371723 on Build GRCh38
7316538 c.5247G>T p.L1749L Chr.9: 111371645-111371645 on Build GRCh38
6693193 c.4695C>T p.T1565T Chr.9: 111371663-111371663 on Build GRCh38
7316536 c.5248G>T p.A1750S Chr.9: 111371644-111371644 on Build GRCh38
4864258 c.4725G>A p.T1575T Chr.9: 111371633-111371633 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766083260 Chr.9: 111371603-111371603 on Build GRCh38
rs369172314 Chr.9: 111371757-111371757 on Build GRCh38
rs1019112115 Chr.9: 111371739-111371739 on Build GRCh38
rs760088981 Chr.9: 111371726-111371726 on Build GRCh38
rs369474247 Chr.9: 111371678-111371678 on Build GRCh38
rs767011360 Chr.9: 111371716-111371716 on Build GRCh38
rs749856712 Chr.9: 111371722-111371722 on Build GRCh38
rs576172285 Chr.9: 111371679-111371679 on Build GRCh38
rs1029435218 Chr.9: 111371720-111371720 on Build GRCh38
rs757167579 Chr.9: 111371743-111371743 on Build GRCh38
rs1022760773 Chr.9: 111371659-111371659 on Build GRCh38
rs765626157 Chr.9: 111371577-111371577 on Build GRCh38
rs770302228 Chr.9: 111371675-111371675 on Build GRCh38
rs889309888 Chr.9: 111371712-111371712 on Build GRCh38
rs1005598325 Chr.9: 111371668-111371668 on Build GRCh38
rs756081876 Chr.9: 111371632-111371632 on Build GRCh38
rs762407676 Chr.9: 111371575-111371575 on Build GRCh38
rs748113907 Chr.9: 111371664-111371664 on Build GRCh38
rs904738572 Chr.9: 111371582-111371582 on Build GRCh38
rs141635106 Chr.9: 111371634-111371634 on Build GRCh38
rs759262864 Chr.9: 111371701-111371701 on Build GRCh38
rs780465272 Chr.9: 111371642-111371642 on Build GRCh38
rs199743974 Chr.9: 111371663-111371663 on Build GRCh38
rs192652307 Chr.9: 111371693-111371693 on Build GRCh38
rs773573276 Chr.9: 111371591-111371591 on Build GRCh38
rs372133962 Chr.9: 111371633-111371633 on Build GRCh38
rs770261631 Chr.9: 111371578-111371579 on Build GRCh38
rs879104288 Chr.9: 111371649-111371649 on Build GRCh38
rs778874758 Chr.9: 111371748-111371748 on Build GRCh38
rs768712662 Chr.9: 111371662-111371662 on Build GRCh38
rs767072601 Chr.9: 111371615-111371615 on Build GRCh38
rs567210589 Chr.9: 111371555-111371555 on Build GRCh38
rs201441042 Chr.9: 111371734-111371734 on Build GRCh38
rs373083332 Chr.9: 111371759-111371759 on Build GRCh38
rs750148961 Chr.9: 111371752-111371752 on Build GRCh38
rs771259015 Chr.9: 111371684-111371684 on Build GRCh38
rs754491585 Chr.9: 111371614-111371614 on Build GRCh38
rs751179387 Chr.9: 111371610-111371610 on Build GRCh38
rs765384754 Chr.9: 111371599-111371599 on Build GRCh38
rs747232413 Chr.9: 111371648-111371648 on Build GRCh38
rs1571978 Chr.9: 111371558-111371558 on Build GRCh38
rs1053673423 Chr.9: 111371609-111371609 on Build GRCh38
rs547211740 Chr.9: 111371554-111371554 on Build GRCh38
rs376101353 Chr.9: 111371738-111371738 on Build GRCh38

View Full Product Details