Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 163326076-163326305 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPY1R
Entrez Gene ID
4886
Gene Name
neuropeptide Y receptor Y1
Gene Aliases
NPY1-R, NPYR
UniGene
Hs.519057
Gene Chromosome Location
Chr.4: 163323962-163344847 on Build GRCh38
Transcripts
NM_000909.5 XM_005263031.3 XM_011532010.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7336776 c.326G>A p.G109D Chr.4: 163326229-163326229 on Build GRCh38
3714737 c.261G>T p.L87F Chr.4: 163326294-163326294 on Build GRCh38
121338 c.436C>A p.R146R Chr.4: 163326119-163326119 on Build GRCh38
3335161 c.312C>G p.D104E Chr.4: 163326243-163326243 on Build GRCh38
5535589 c.477T>A p.I159I Chr.4: 163326078-163326078 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765083532 Chr.4: 163326270-163326270 on Build GRCh38
rs867611414 Chr.4: 163326206-163326206 on Build GRCh38
rs756536910 Chr.4: 163326186-163326186 on Build GRCh38
rs78156188 Chr.4: 163326223-163326223 on Build GRCh38
rs368406189 Chr.4: 163326258-163326258 on Build GRCh38
rs750120094 Chr.4: 163326132-163326132 on Build GRCh38
rs748609830 Chr.4: 163326185-163326185 on Build GRCh38
rs201300913 Chr.4: 163326201-163326201 on Build GRCh38
rs375737343 Chr.4: 163326222-163326222 on Build GRCh38
rs757920246 Chr.4: 163326140-163326140 on Build GRCh38
rs764890620 Chr.4: 163326124-163326124 on Build GRCh38
rs926086441 Chr.4: 163326272-163326272 on Build GRCh38
rs768758335 Chr.4: 163326225-163326225 on Build GRCh38
rs148479836 Chr.4: 163326118-163326118 on Build GRCh38
rs755519745 Chr.4: 163326163-163326163 on Build GRCh38
rs936080593 Chr.4: 163326284-163326284 on Build GRCh38
rs183843859 Chr.4: 163326119-163326119 on Build GRCh38
rs751066628 Chr.4: 163326156-163326156 on Build GRCh38
rs771042799 Chr.4: 163326221-163326221 on Build GRCh38
rs781648360 Chr.4: 163326174-163326174 on Build GRCh38
rs142702739 Chr.4: 163326155-163326155 on Build GRCh38

View Full Product Details