Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90980825-90981096 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRC1
Entrez Gene ID
9055
Gene Name
protein regulator of cytokinesis 1
Gene Aliases
ASE1
UniGene
Hs.366401
Gene Chromosome Location
Chr.15: 90966038-90994651 on Build GRCh38
Transcripts
NM_001267580.1 NM_003981.3 NM_199413.2 XM_005254987.2 XM_006720759.1 XM_006720760.1 XM_011522187.1 XM_011522188.2 XM_011522189.1 XM_011522190.2 XM_011522191.2 XM_011522192.1 XM_017022712.1 XM_017022713.1 XM_017022714.1 XM_017022715.1 XM_017022716.1 XM_017022717.1 NR_051984.1

Gene Symbol
PRC1-AS1
Entrez Gene ID
100507118
Gene Name
PRC1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.366401
Gene Chromosome Location
Chr.15: 90966369-90988624 on Build GRCh38
Transcripts
NM_001267580.1 NM_003981.3 NM_199413.2 XM_005254987.2 XM_006720759.1 XM_006720760.1 XM_011522187.1 XM_011522188.2 XM_011522189.1 XM_011522190.2 XM_011522191.2 XM_011522192.1 XM_017022712.1 XM_017022713.1 XM_017022714.1 XM_017022715.1 XM_017022716.1 XM_017022717.1 NR_051984.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6021378 c.703G>C p.V235L Chr.15: 90981003-90981003 on Build GRCh38
6538391 c.675G>C p.L225L Chr.15: 90981031-90981031 on Build GRCh38
5363163 c.787A>G p.I263V Chr.15: 90980919-90980919 on Build GRCh38
4057978 c.731G>A p.R244Q Chr.15: 90980975-90980975 on Build GRCh38
1196581 c.781G>C p.A261P Chr.15: 90980925-90980925 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761956294 Chr.15: 90981013-90981013 on Build GRCh38
rs780486994 Chr.15: 90980930-90980930 on Build GRCh38
rs750676488 Chr.15: 90981026-90981026 on Build GRCh38
rs200799126 Chr.15: 90980943-90980943 on Build GRCh38
rs200688029 Chr.15: 90980873-90980873 on Build GRCh38
rs974862128 Chr.15: 90980959-90980959 on Build GRCh38
rs966413246 Chr.15: 90981049-90981049 on Build GRCh38
rs767594265 Chr.15: 90981015-90981015 on Build GRCh38
rs533610719 Chr.15: 90981005-90981005 on Build GRCh38
rs953302509 Chr.15: 90980923-90980924 on Build GRCh38
rs771644143 Chr.15: 90981074-90981074 on Build GRCh38
rs142300202 Chr.15: 90980892-90980892 on Build GRCh38
rs548375030 Chr.15: 90981075-90981075 on Build GRCh38
rs776619186 Chr.15: 90980988-90980988 on Build GRCh38
rs779491619 Chr.15: 90981062-90981062 on Build GRCh38
rs895226053 Chr.15: 90981064-90981064 on Build GRCh38
rs149143720 Chr.15: 90980891-90980891 on Build GRCh38
rs1004888334 Chr.15: 90981039-90981039 on Build GRCh38
rs780311578 Chr.15: 90981003-90981003 on Build GRCh38
rs760535884 Chr.15: 90980885-90980885 on Build GRCh38
rs769188156 Chr.15: 90980966-90980966 on Build GRCh38
rs755467130 Chr.15: 90981056-90981056 on Build GRCh38
rs200105515 Chr.15: 90980920-90980920 on Build GRCh38
rs767270485 Chr.15: 90980876-90980876 on Build GRCh38
rs764248316 Chr.15: 90980894-90980894 on Build GRCh38
rs767805068 Chr.15: 90980933-90980933 on Build GRCh38
rs759517630 Chr.15: 90981003-90981003 on Build GRCh38
rs751534073 Chr.15: 90981010-90981010 on Build GRCh38
rs781489628 Chr.15: 90980904-90980904 on Build GRCh38
rs770872351 Chr.15: 90980987-90980987 on Build GRCh38
rs761339215 Chr.15: 90980870-90980870 on Build GRCh38
rs760507368 Chr.15: 90980984-90980984 on Build GRCh38
rs199719104 Chr.15: 90980858-90980858 on Build GRCh38
rs547936544 Chr.15: 90980978-90980978 on Build GRCh38
rs777511352 Chr.15: 90980859-90980860 on Build GRCh38
rs149838434 Chr.15: 90980979-90980979 on Build GRCh38
rs375542624 Chr.15: 90980884-90980884 on Build GRCh38
rs371959663 Chr.15: 90981042-90981042 on Build GRCh38
rs529948276 Chr.15: 90980935-90980935 on Build GRCh38
rs758465836 Chr.15: 90980936-90980940 on Build GRCh38
rs368282015 Chr.15: 90981037-90981037 on Build GRCh38
rs745950649 Chr.15: 90980924-90980929 on Build GRCh38
rs772926435 Chr.15: 90980980-90980980 on Build GRCh38
rs778285093 Chr.15: 90980975-90980975 on Build GRCh38
rs151276157 Chr.15: 90980897-90980897 on Build GRCh38
rs79586460 Chr.15: 90981052-90981052 on Build GRCh38
rs201945279 Chr.15: 90981063-90981063 on Build GRCh38
rs749769477 Chr.15: 90980964-90980964 on Build GRCh38
rs750686979 Chr.15: 90980908-90980908 on Build GRCh38
rs372796823 Chr.15: 90980857-90980857 on Build GRCh38
rs747209200 Chr.15: 90981076-90981077 on Build GRCh38

View Full Product Details