Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33290243-33290478 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR6834
Entrez Gene ID
102465501
Gene Name
microRNA 6834
Gene Aliases
hsa-mir-6834
UniGene
Gene Chromosome Location
Chr.6: 33290245-33290325 on Build GRCh38
Transcripts
NR_106892.1 NM_001185181.2 NM_001265595.1 NM_001265596.1 NM_014260.3

Gene Symbol
PFDN6
Entrez Gene ID
10471
Gene Name
prefoldin subunit 6
Gene Aliases
H2-KE2, HKE2, KE-2, PFD6
UniGene
Hs.446374
Gene Chromosome Location
Chr.6: 33289597-33290934 on Build GRCh38
Transcripts
NR_106892.1 NM_001185181.2 NM_001265595.1 NM_001265596.1 NM_014260.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5202234 c.160A>G p.N54D Chr.6: 33290350-33290350 on Build GRCh38
6379511 c.235A>G p.R79G Chr.6: 33290425-33290425 on Build GRCh38
5208475 c.234G>A p.K78K Chr.6: 33290424-33290424 on Build GRCh38
5459250 c.218G>A p.R73Q Chr.6: 33290408-33290408 on Build GRCh38
451300 c.160A>G p.N54D Chr.6: 33290350-33290350 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757861907 Chr.6: 33290323-33290323 on Build GRCh38
rs533672416 Chr.6: 33290346-33290346 on Build GRCh38
rs372709557 Chr.6: 33290458-33290458 on Build GRCh38
rs566658142 Chr.6: 33290304-33290304 on Build GRCh38
rs761112243 Chr.6: 33290294-33290294 on Build GRCh38
rs147946254 Chr.6: 33290395-33290395 on Build GRCh38
rs1049515164 Chr.6: 33290337-33290337 on Build GRCh38
rs772219198 Chr.6: 33290435-33290435 on Build GRCh38
rs112260262 Chr.6: 33290451-33290451 on Build GRCh38
rs376399387 Chr.6: 33290368-33290368 on Build GRCh38
rs373417699 Chr.6: 33290266-33290266 on Build GRCh38
rs748190140 Chr.6: 33290284-33290284 on Build GRCh38
rs370401018 Chr.6: 33290415-33290415 on Build GRCh38
rs772455449 Chr.6: 33290289-33290289 on Build GRCh38
rs752222785 Chr.6: 33290319-33290319 on Build GRCh38
rs113189332 Chr.6: 33290318-33290318 on Build GRCh38
rs773504095 Chr.6: 33290290-33290290 on Build GRCh38
rs748245009 Chr.6: 33290420-33290420 on Build GRCh38
rs181437986 Chr.6: 33290279-33290279 on Build GRCh38
rs764772400 Chr.6: 33290306-33290306 on Build GRCh38
rs557710967 Chr.6: 33290295-33290295 on Build GRCh38
rs763752325 Chr.6: 33290345-33290345 on Build GRCh38
rs1008084563 Chr.6: 33290398-33290398 on Build GRCh38
rs985139018 Chr.6: 33290408-33290408 on Build GRCh38
rs142890625 Chr.6: 33290391-33290391 on Build GRCh38
rs780823244 Chr.6: 33290353-33290353 on Build GRCh38

View Full Product Details