Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 30910844-30911117 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT14
Entrez Gene ID
79623
Gene Name
polypeptide N-acetylgalactosaminyltransferase 14
Gene Aliases
GALNT15, GalNac-T10, GalNac-T14
UniGene
Hs.468058
Gene Chromosome Location
Chr.2: 30888518-31138726 on Build GRCh38
Transcripts
NM_001253826.1 NM_001253827.1 NM_024572.3 XM_011533105.1 XM_017004906.1 XM_017004907.1 XM_017004908.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6669026 c.1644C>T p.D548D Chr.2: 30910916-30910916 on Build GRCh38
1020075 c.1587C>T p.N529N Chr.2: 30910973-30910973 on Build GRCh38
1020076 c.1515T>G p.T505T Chr.2: 30911045-30911045 on Build GRCh38
5043980 c.1603G>C p.V535L Chr.2: 30910957-30910957 on Build GRCh38
6542486 c.1584G>A p.E528E Chr.2: 30910976-30910976 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202186542 Chr.2: 30910957-30910957 on Build GRCh38
rs777221227 Chr.2: 30910929-30910929 on Build GRCh38
rs776911563 Chr.2: 30910871-30910871 on Build GRCh38
rs754936068 Chr.2: 30910912-30910912 on Build GRCh38
rs866967833 Chr.2: 30910914-30910914 on Build GRCh38
rs763186258 Chr.2: 30910897-30910897 on Build GRCh38
rs749371562 Chr.2: 30911000-30911000 on Build GRCh38
rs551834022 Chr.2: 30910960-30910960 on Build GRCh38
rs772350834 Chr.2: 30910863-30910863 on Build GRCh38
rs868796616 Chr.2: 30910946-30910946 on Build GRCh38
rs150328877 Chr.2: 30911005-30911005 on Build GRCh38
rs372565159 Chr.2: 30911026-30911026 on Build GRCh38
rs772181235 Chr.2: 30910872-30910873 on Build GRCh38
rs976965495 Chr.2: 30911078-30911078 on Build GRCh38
rs373341564 Chr.2: 30911080-30911080 on Build GRCh38
rs778608668 Chr.2: 30910980-30910980 on Build GRCh38
rs756871233 Chr.2: 30910931-30910931 on Build GRCh38
rs775870653 Chr.2: 30910866-30910866 on Build GRCh38
rs373001089 Chr.2: 30910933-30910933 on Build GRCh38
rs749365568 Chr.2: 30911093-30911093 on Build GRCh38
rs770358878 Chr.2: 30910991-30910991 on Build GRCh38
rs759633341 Chr.2: 30911007-30911007 on Build GRCh38
rs767426919 Chr.2: 30910921-30910921 on Build GRCh38
rs761577729 Chr.2: 30910875-30910875 on Build GRCh38
rs771872193 Chr.2: 30911015-30911015 on Build GRCh38
rs758066912 Chr.2: 30911038-30911038 on Build GRCh38
rs759350736 Chr.2: 30910966-30910966 on Build GRCh38
rs761963570 Chr.2: 30910884-30910884 on Build GRCh38
rs369746576 Chr.2: 30910982-30910982 on Build GRCh38
rs756741132 Chr.2: 30911068-30911068 on Build GRCh38
rs761460016 Chr.2: 30910896-30910897 on Build GRCh38
rs755587020 Chr.2: 30910923-30910923 on Build GRCh38
rs376941946 Chr.2: 30911057-30911057 on Build GRCh38
rs748627210 Chr.2: 30910955-30910955 on Build GRCh38
rs774070861 Chr.2: 30910961-30910961 on Build GRCh38
rs760676083 Chr.2: 30911021-30911021 on Build GRCh38
rs973285495 Chr.2: 30911032-30911032 on Build GRCh38
rs774598779 Chr.2: 30911006-30911006 on Build GRCh38
rs753387439 Chr.2: 30910978-30910978 on Build GRCh38
rs1039199493 Chr.2: 30910876-30910876 on Build GRCh38
rs775203378 Chr.2: 30911020-30911020 on Build GRCh38
rs200522917 Chr.2: 30910983-30910983 on Build GRCh38
rs777248765 Chr.2: 30910953-30910953 on Build GRCh38
rs577646417 Chr.2: 30910932-30910932 on Build GRCh38
rs2278080 Chr.2: 30910896-30910896 on Build GRCh38
rs766100424 Chr.2: 30910900-30910900 on Build GRCh38
rs766040667 Chr.2: 30911093-30911094 on Build GRCh38
rs765292501 Chr.2: 30911022-30911022 on Build GRCh38
rs565883379 Chr.2: 30911033-30911033 on Build GRCh38
rs2278079 Chr.2: 30911073-30911073 on Build GRCh38
rs145219116 Chr.2: 30910973-30910973 on Build GRCh38
rs753339938 Chr.2: 30911030-30911030 on Build GRCh38
rs756921306 Chr.2: 30910979-30910979 on Build GRCh38
rs751300380 Chr.2: 30910906-30910906 on Build GRCh38
rs752251096 Chr.2: 30910922-30910922 on Build GRCh38
rs747479883 Chr.2: 30910869-30910869 on Build GRCh38
rs769094837 Chr.2: 30910936-30910936 on Build GRCh38
rs151144957 Chr.2: 30910990-30910990 on Build GRCh38
rs750350211 Chr.2: 30911071-30911073 on Build GRCh38
rs201589708 Chr.2: 30910972-30910972 on Build GRCh38
rs781693188 Chr.2: 30911040-30911040 on Build GRCh38
rs770016745 Chr.2: 30910956-30910956 on Build GRCh38
rs759862969 Chr.2: 30910867-30910868 on Build GRCh38
rs981927114 Chr.2: 30911003-30911003 on Build GRCh38
rs764008117 Chr.2: 30911023-30911023 on Build GRCh38
rs762958687 Chr.2: 30911074-30911075 on Build GRCh38
rs776493492 Chr.2: 30910894-30910894 on Build GRCh38
rs769204797 Chr.2: 30910870-30910870 on Build GRCh38
rs763603221 Chr.2: 30910977-30910977 on Build GRCh38
rs767480097 Chr.2: 30910969-30910969 on Build GRCh38

View Full Product Details