Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 7825727-7826000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR5P3
Entrez Gene ID
120066
Gene Name
olfactory receptor family 5 subfamily P member 3
Gene Aliases
JCG1
UniGene
Hs.351825
Gene Chromosome Location
Chr.11: 7825037-7825972 on Build GRCh38
Transcripts
NM_153445.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6993885 c.227C>T p.S76L Chr.11: 7825746-7825746 on Build GRCh38
3870292 c.40C>T p.L14F Chr.11: 7825933-7825933 on Build GRCh38
2019939 c.153_155delAAG p.R51delR Chr.11: 7825818-7825820 on Build GRCh38
2019937 c.215G>A p.G72E Chr.11: 7825758-7825758 on Build GRCh38
3967860 c.239T>C p.V80A Chr.11: 7825734-7825734 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1017139600 Chr.11: 7825825-7825825 on Build GRCh38
rs777837463 Chr.11: 7825776-7825776 on Build GRCh38
rs907357450 Chr.11: 7825948-7825948 on Build GRCh38
rs774154347 Chr.11: 7825782-7825782 on Build GRCh38
rs145234842 Chr.11: 7825818-7825820 on Build GRCh38
rs755748214 Chr.11: 7825765-7825765 on Build GRCh38
rs116328989 Chr.11: 7825831-7825831 on Build GRCh38
rs1482793 Chr.11: 7825925-7825925 on Build GRCh38
rs763859768 Chr.11: 7825952-7825952 on Build GRCh38
rs753330127 Chr.11: 7825955-7825955 on Build GRCh38
rs752364154 Chr.11: 7825940-7825940 on Build GRCh38
rs536977656 Chr.11: 7825975-7825975 on Build GRCh38
rs267603206 Chr.11: 7825933-7825933 on Build GRCh38
rs754613368 Chr.11: 7825757-7825757 on Build GRCh38
rs755660443 Chr.11: 7825862-7825862 on Build GRCh38
rs757168362 Chr.11: 7825873-7825873 on Build GRCh38
rs775755921 Chr.11: 7825800-7825800 on Build GRCh38
rs780233932 Chr.11: 7825907-7825907 on Build GRCh38
rs750787360 Chr.11: 7825752-7825752 on Build GRCh38
rs61742097 Chr.11: 7825911-7825911 on Build GRCh38
rs745672119 Chr.11: 7825896-7825896 on Build GRCh38
rs760362208 Chr.11: 7825943-7825943 on Build GRCh38
rs767270354 Chr.11: 7825859-7825859 on Build GRCh38
rs781009037 Chr.11: 7825764-7825764 on Build GRCh38
rs763359420 Chr.11: 7825930-7825930 on Build GRCh38
rs776646418 Chr.11: 7825914-7825914 on Build GRCh38
rs777463642 Chr.11: 7825867-7825867 on Build GRCh38
rs753319106 Chr.11: 7825872-7825872 on Build GRCh38
rs369784933 Chr.11: 7825837-7825837 on Build GRCh38
rs533589718 Chr.11: 7825826-7825826 on Build GRCh38
rs761815839 Chr.11: 7825918-7825918 on Build GRCh38
rs544874756 Chr.11: 7825789-7825789 on Build GRCh38
rs750745773 Chr.11: 7825836-7825836 on Build GRCh38
rs746831577 Chr.11: 7825978-7825978 on Build GRCh38
rs1482792 Chr.11: 7825919-7825919 on Build GRCh38
rs749294640 Chr.11: 7825779-7825779 on Build GRCh38
rs771913202 Chr.11: 7825904-7825904 on Build GRCh38
rs199997958 Chr.11: 7825766-7825766 on Build GRCh38
rs769707514 Chr.11: 7825876-7825876 on Build GRCh38
rs756724882 Chr.11: 7825965-7825965 on Build GRCh38
rs60467657 Chr.11: 7825801-7825801 on Build GRCh38
rs752452303 Chr.11: 7825860-7825860 on Build GRCh38
rs768899891 Chr.11: 7825913-7825913 on Build GRCh38
rs372277097 Chr.11: 7825892-7825892 on Build GRCh38
rs762344051 Chr.11: 7825824-7825824 on Build GRCh38
rs764208755 Chr.11: 7825804-7825804 on Build GRCh38
rs1482794 Chr.11: 7825976-7825976 on Build GRCh38
rs778896902 Chr.11: 7825970-7825970 on Build GRCh38
rs766896087 Chr.11: 7825937-7825937 on Build GRCh38
rs770599210 Chr.11: 7825780-7825780 on Build GRCh38
rs772279725 Chr.11: 7825797-7825797 on Build GRCh38
rs777092731 Chr.11: 7825806-7825806 on Build GRCh38

View Full Product Details