Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 124746683-124746924 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR37
Entrez Gene ID
2861
Gene Name
G protein-coupled receptor 37
Gene Aliases
EDNRBL, PAELR, hET(B)R-LP
UniGene
Hs.406094
Gene Chromosome Location
Chr.7: 124745601-124766025 on Build GRCh38
Transcripts
NM_005302.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1312632 c.1453C>A p.Q485K Chr.7: 124746914-124746914 on Build GRCh38
5910571 c.1522G>A p.E508K Chr.7: 124746845-124746845 on Build GRCh38
7370050 c.1551C>G p.Y517* Chr.7: 124746816-124746816 on Build GRCh38
4978225 c.1461G>C p.E487D Chr.7: 124746906-124746906 on Build GRCh38
1239015 c.1445G>A p.R482Q Chr.7: 124746922-124746922 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530322862 Chr.7: 124746888-124746888 on Build GRCh38
rs757458367 Chr.7: 124746756-124746756 on Build GRCh38
rs771955298 Chr.7: 124746723-124746723 on Build GRCh38
rs775441914 Chr.7: 124746894-124746894 on Build GRCh38
rs778319928 Chr.7: 124746831-124746831 on Build GRCh38
rs753480808 Chr.7: 124746817-124746817 on Build GRCh38
rs143006442 Chr.7: 124746761-124746761 on Build GRCh38
rs758425049 Chr.7: 124746848-124746848 on Build GRCh38
rs748050700 Chr.7: 124746842-124746842 on Build GRCh38
rs765285440 Chr.7: 124746788-124746788 on Build GRCh38
rs567822901 Chr.7: 124746864-124746864 on Build GRCh38
rs777647717 Chr.7: 124746858-124746858 on Build GRCh38
rs367965158 Chr.7: 124746804-124746804 on Build GRCh38
rs759651070 Chr.7: 124746784-124746784 on Build GRCh38
rs976904661 Chr.7: 124746750-124746750 on Build GRCh38
rs111247791 Chr.7: 124746895-124746895 on Build GRCh38
rs754480987 Chr.7: 124746820-124746820 on Build GRCh38
rs748138558 Chr.7: 124746708-124746708 on Build GRCh38
rs746105366 Chr.7: 124746889-124746889 on Build GRCh38
rs910630336 Chr.7: 124746851-124746851 on Build GRCh38
rs545769416 Chr.7: 124746729-124746729 on Build GRCh38
rs183803404 Chr.7: 124746806-124746806 on Build GRCh38
rs980429686 Chr.7: 124746715-124746715 on Build GRCh38
rs773164296 Chr.7: 124746735-124746735 on Build GRCh38
rs776789829 Chr.7: 124746762-124746762 on Build GRCh38
rs753205316 Chr.7: 124746798-124746798 on Build GRCh38
rs142854495 Chr.7: 124746892-124746892 on Build GRCh38
rs770691420 Chr.7: 124746885-124746885 on Build GRCh38
rs551314420 Chr.7: 124746814-124746814 on Build GRCh38
rs972359681 Chr.7: 124746783-124746783 on Build GRCh38
rs757684396 Chr.7: 124746808-124746808 on Build GRCh38
rs150466939 Chr.7: 124746741-124746741 on Build GRCh38

View Full Product Details