Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219505989-219506248 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMPPA
Entrez Gene ID
29926
Gene Name
GDP-mannose pyrophosphorylase A
Gene Aliases
AAMR
UniGene
Hs.27059
Gene Chromosome Location
Chr.2: 219498865-219506996 on Build GRCh38
Transcripts
NM_013335.3 NM_205847.2 XM_005246483.3 XM_005246486.3 XM_011511032.2 XM_017003906.1 XM_017003907.1 XM_017003908.1 XM_017003909.1 XM_017003910.1 XM_017003911.1 XM_017003912.1 XM_017003913.1 XM_017003914.1 XM_017003915.1 XM_017003916.1 XM_017003917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4800949 c.914T>C p.V305A Chr.2: 219505993-219505993 on Build GRCh38
1253345 c.936C>T p.T312T Chr.2: 219506015-219506015 on Build GRCh38
6859154 c.969C>T p.I323I Chr.2: 219506048-219506048 on Build GRCh38
3578043 c.918C>T p.S306S Chr.2: 219505997-219505997 on Build GRCh38
1016825 c.910A>G p.N304D Chr.2: 219505989-219505989 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs145626765 Chr.2: 219506184-219506184 on Build GRCh38
rs907394838 Chr.2: 219506183-219506183 on Build GRCh38
rs889780954 Chr.2: 219506129-219506129 on Build GRCh38
rs1024873807 Chr.2: 219506176-219506176 on Build GRCh38
rs193268245 Chr.2: 219506105-219506105 on Build GRCh38
rs764413927 Chr.2: 219506031-219506031 on Build GRCh38
rs1016890574 Chr.2: 219506132-219506132 on Build GRCh38
rs552239387 Chr.2: 219506133-219506133 on Build GRCh38
rs746084181 Chr.2: 219506084-219506084 on Build GRCh38
rs765253658 Chr.2: 219506208-219506208 on Build GRCh38
rs370554538 Chr.2: 219506225-219506225 on Build GRCh38
rs373367877 Chr.2: 219506118-219506118 on Build GRCh38
rs749701763 Chr.2: 219506102-219506102 on Build GRCh38
rs989119234 Chr.2: 219506200-219506200 on Build GRCh38
rs754888360 Chr.2: 219506207-219506207 on Build GRCh38
rs781419973 Chr.2: 219506070-219506070 on Build GRCh38
rs549821547 Chr.2: 219506037-219506037 on Build GRCh38
rs753658708 Chr.2: 219506122-219506122 on Build GRCh38
rs755878658 Chr.2: 219506029-219506029 on Build GRCh38
rs779038410 Chr.2: 219506038-219506038 on Build GRCh38
rs199578533 Chr.2: 219506196-219506196 on Build GRCh38
rs914961808 Chr.2: 219506040-219506040 on Build GRCh38
rs565878918 Chr.2: 219506162-219506162 on Build GRCh38
rs13428617 Chr.2: 219506024-219506024 on Build GRCh38
rs994280323 Chr.2: 219506169-219506169 on Build GRCh38
rs972619868 Chr.2: 219506137-219506137 on Build GRCh38
rs753854673 Chr.2: 219506086-219506086 on Build GRCh38
rs997489319 Chr.2: 219506090-219506090 on Build GRCh38
rs376825523 Chr.2: 219506015-219506015 on Build GRCh38
rs945160946 Chr.2: 219506048-219506048 on Build GRCh38
rs532396940 Chr.2: 219506119-219506119 on Build GRCh38
rs757296974 Chr.2: 219506224-219506224 on Build GRCh38
rs541265208 Chr.2: 219506007-219506007 on Build GRCh38
rs897216358 Chr.2: 219506165-219506165 on Build GRCh38
rs747082001 Chr.2: 219506039-219506039 on Build GRCh38
rs757415529 Chr.2: 219506141-219506141 on Build GRCh38
rs960504026 Chr.2: 219506156-219506156 on Build GRCh38
rs772851876 Chr.2: 219506108-219506108 on Build GRCh38
rs1025784586 Chr.2: 219506144-219506144 on Build GRCh38
rs759003531 Chr.2: 219506199-219506199 on Build GRCh38
rs376462993 Chr.2: 219506103-219506103 on Build GRCh38
rs149492263 Chr.2: 219506032-219506032 on Build GRCh38
rs370957725 Chr.2: 219506016-219506016 on Build GRCh38
rs376516329 Chr.2: 219506214-219506214 on Build GRCh38
rs775853885 Chr.2: 219506100-219506100 on Build GRCh38
rs563311663 Chr.2: 219506054-219506054 on Build GRCh38

View Full Product Details