Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129111498-129111743 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPA
Entrez Gene ID
5524
Gene Name
protein phosphatase 2 phosphatase activator
Gene Aliases
PP2A, PPP2R4, PR53
UniGene
Hs.400740
Gene Chromosome Location
Chr.9: 129110949-129148946 on Build GRCh38
Transcripts
NM_001193397.1 NM_001271832.1 NM_021131.4 NM_178000.2 NM_178001.2 NM_178003.2 XM_011518834.2 XM_011518835.2 XM_011518836.2 XM_011518837.2 XM_011518838.2 XM_017014888.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5410513 c.10G>A p.G4S Chr.9: 129111610-129111610 on Build GRCh38
5410512 c.9G>A p.E3E Chr.9: 129111609-129111609 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs888344325 Chr.9: 129111704-129111704 on Build GRCh38
rs941824644 Chr.9: 129111722-129111722 on Build GRCh38
rs532691946 Chr.9: 129111533-129111533 on Build GRCh38
rs537134632 Chr.9: 129111682-129111682 on Build GRCh38
rs965353380 Chr.9: 129111596-129111596 on Build GRCh38
rs1018324884 Chr.9: 129111593-129111593 on Build GRCh38
rs910033726 Chr.9: 129111688-129111688 on Build GRCh38
rs60764362 Chr.9: 129111606-129111606 on Build GRCh38
rs945106750 Chr.9: 129111621-129111621 on Build GRCh38
rs534864957 Chr.9: 129111638-129111638 on Build GRCh38
rs557030435 Chr.9: 129111689-129111689 on Build GRCh38
rs552566278 Chr.9: 129111575-129111575 on Build GRCh38
rs773502942 Chr.9: 129111624-129111624 on Build GRCh38
rs368782227 Chr.9: 129111586-129111586 on Build GRCh38
rs1049684836 Chr.9: 129111695-129111695 on Build GRCh38
rs554650635 Chr.9: 129111639-129111639 on Build GRCh38
rs568208661 Chr.9: 129111663-129111663 on Build GRCh38
rs770962604 Chr.9: 129111647-129111647 on Build GRCh38
rs986569137 Chr.9: 129111673-129111673 on Build GRCh38
rs747138324 Chr.9: 129111635-129111648 on Build GRCh38
rs936199816 Chr.9: 129111554-129111555 on Build GRCh38
rs955312259 Chr.9: 129111668-129111668 on Build GRCh38

View Full Product Details