Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44006332-44006577 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAGS
Entrez Gene ID
162417
Gene Name
N-acetylglutamate synthase
Gene Aliases
AGAS, ARGA
UniGene
Hs.8876
Gene Chromosome Location
Chr.17: 44004546-44009068 on Build GRCh38
Transcripts
NM_153006.2 XM_011524438.1 XM_011524439.2 XM_011525035.1

Gene Symbol
PYY
Entrez Gene ID
5697
Gene Name
peptide YY
Gene Aliases
PYY-I, PYY1
UniGene
Hs.169249
Gene Chromosome Location
Chr.17: 43952733-44024260 on Build GRCh38
Transcripts
NM_153006.2 XM_011524438.1 XM_011524439.2 XM_011525035.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6016671 c.944A>T p.D315V Chr.17: 44006557-44006557 on Build GRCh38
6377639 c.956T>C p.V319A Chr.17: 44006569-44006569 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs754360456 Chr.17: 44006421-44006421 on Build GRCh38
rs952633263 Chr.17: 44006507-44006507 on Build GRCh38
rs780597835 Chr.17: 44006549-44006549 on Build GRCh38
rs186127828 Chr.17: 44006553-44006553 on Build GRCh38
rs1000293040 Chr.17: 44006498-44006498 on Build GRCh38
rs730880267 Chr.17: 44006527-44006527 on Build GRCh38
rs749692636 Chr.17: 44006552-44006552 on Build GRCh38
rs370356326 Chr.17: 44006556-44006556 on Build GRCh38
rs550798774 Chr.17: 44006469-44006469 on Build GRCh38
rs183275808 Chr.17: 44006490-44006490 on Build GRCh38
rs947581783 Chr.17: 44006401-44006401 on Build GRCh38
rs551054997 Chr.17: 44006540-44006540 on Build GRCh38
rs764590560 Chr.17: 44006492-44006492 on Build GRCh38
rs758117966 Chr.17: 44006504-44006504 on Build GRCh38
rs376106132 Chr.17: 44006491-44006491 on Build GRCh38
rs760506594 Chr.17: 44006478-44006478 on Build GRCh38
rs752307842 Chr.17: 44006496-44006496 on Build GRCh38
rs1047973516 Chr.17: 44006445-44006445 on Build GRCh38
rs763878126 Chr.17: 44006529-44006529 on Build GRCh38
rs896090140 Chr.17: 44006452-44006453 on Build GRCh38
rs1023579094 Chr.17: 44006476-44006476 on Build GRCh38
rs756497810 Chr.17: 44006548-44006548 on Build GRCh38
rs903384515 Chr.17: 44006444-44006444 on Build GRCh38
rs228770 Chr.17: 44006472-44006472 on Build GRCh38
rs1041987739 Chr.17: 44006428-44006428 on Build GRCh38

View Full Product Details