Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 97686587-97686834 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTDH
Entrez Gene ID
92140
Gene Name
metadherin
Gene Aliases
3D3, AEG-1, AEG1, LYRIC, LYRIC/3D3
UniGene
Hs.377155
Gene Chromosome Location
Chr.8: 97643972-97730260 on Build GRCh38
Transcripts
NM_178812.3 XM_005251099.3 XM_011517367.2 XM_011517368.2 XM_011517369.2 XM_011517370.2 XM_017013966.1 XM_017013967.1 XM_017013968.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6472767 c.539C>A p.S180* Chr.8: 97686723-97686723 on Build GRCh38
1102806 c.542G>T p.R181L Chr.8: 97686726-97686726 on Build GRCh38
7242250 c.528G>A p.V176V Chr.8: 97686712-97686712 on Build GRCh38
6472766 c.538T>G p.S180A Chr.8: 97686722-97686722 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs997160188 Chr.8: 97686699-97686699 on Build GRCh38
rs761455692 Chr.8: 97686704-97686704 on Build GRCh38
rs768407816 Chr.8: 97686655-97686655 on Build GRCh38
rs778887591 Chr.8: 97686629-97686629 on Build GRCh38
rs562197979 Chr.8: 97686777-97686777 on Build GRCh38
rs775014678 Chr.8: 97686738-97686738 on Build GRCh38
rs1128828 Chr.8: 97686743-97686743 on Build GRCh38
rs753856978 Chr.8: 97686785-97686785 on Build GRCh38
rs753146132 Chr.8: 97686761-97686761 on Build GRCh38
rs771865825 Chr.8: 97686641-97686641 on Build GRCh38
rs372330645 Chr.8: 97686772-97686772 on Build GRCh38
rs752994311 Chr.8: 97686793-97686793 on Build GRCh38
rs368949019 Chr.8: 97686739-97686739 on Build GRCh38
rs775995787 Chr.8: 97686661-97686661 on Build GRCh38
rs190066143 Chr.8: 97686756-97686756 on Build GRCh38
rs757332105 Chr.8: 97686793-97686793 on Build GRCh38
rs374847603 Chr.8: 97686725-97686725 on Build GRCh38
rs200266145 Chr.8: 97686719-97686719 on Build GRCh38
rs201472529 Chr.8: 97686706-97686706 on Build GRCh38
rs778762844 Chr.8: 97686794-97686794 on Build GRCh38
rs745593531 Chr.8: 97686640-97686640 on Build GRCh38
rs377400535 Chr.8: 97686619-97686619 on Build GRCh38
rs779704792 Chr.8: 97686643-97686643 on Build GRCh38
rs746668524 Chr.8: 97686653-97686653 on Build GRCh38
rs375438148 Chr.8: 97686801-97686801 on Build GRCh38
rs200623302 Chr.8: 97686784-97686784 on Build GRCh38

View Full Product Details