Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 65704783-65705040 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTFR1
Entrez Gene ID
9650
Gene Name
mitochondrial fission regulator 1
Gene Aliases
CHPPR, FAM54A2
UniGene
Hs.584788
Gene Chromosome Location
Chr.8: 65643873-65771252 on Build GRCh38
Transcripts
NM_001145838.1 NM_001145839.1 NM_014637.3 XM_006716484.2 XM_011517626.1 XM_011517627.2 XM_011517628.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2720669 c.403C>T p.L135L Chr.8: 65704815-65704815 on Build GRCh38
1457820 c.492G>A p.Q164Q Chr.8: 65704904-65704904 on Build GRCh38
5479715 c.373C>G p.Q125E Chr.8: 65704785-65704785 on Build GRCh38
3996101 c.480A>C p.K160N Chr.8: 65704892-65704892 on Build GRCh38
7261086 c.435C>T p.C145C Chr.8: 65704847-65704847 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148450715 Chr.8: 65704874-65704874 on Build GRCh38
rs759569495 Chr.8: 65704930-65704930 on Build GRCh38
rs948871470 Chr.8: 65704977-65704977 on Build GRCh38
rs758264647 Chr.8: 65704823-65704823 on Build GRCh38
rs777075566 Chr.8: 65704902-65704902 on Build GRCh38
rs766314303 Chr.8: 65704834-65704834 on Build GRCh38
rs373638323 Chr.8: 65704935-65704935 on Build GRCh38
rs376894932 Chr.8: 65704906-65704906 on Build GRCh38
rs749451702 Chr.8: 65704850-65704850 on Build GRCh38
rs764579651 Chr.8: 65704806-65704806 on Build GRCh38
rs761574104 Chr.8: 65704813-65704813 on Build GRCh38
rs776145119 Chr.8: 65704807-65704807 on Build GRCh38
rs751417558 Chr.8: 65704927-65704927 on Build GRCh38
rs150643671 Chr.8: 65704905-65704905 on Build GRCh38
rs966369731 Chr.8: 65704859-65704859 on Build GRCh38
rs73240744 Chr.8: 65705000-65705000 on Build GRCh38
rs183603692 Chr.8: 65705004-65705004 on Build GRCh38
rs552169874 Chr.8: 65704918-65704918 on Build GRCh38
rs1057255532 Chr.8: 65704944-65704944 on Build GRCh38
rs17853560 Chr.8: 65704816-65704816 on Build GRCh38
rs149744161 Chr.8: 65704847-65704847 on Build GRCh38
rs144568946 Chr.8: 65704853-65704853 on Build GRCh38
rs894522648 Chr.8: 65704954-65704954 on Build GRCh38
rs201556776 Chr.8: 65704841-65704841 on Build GRCh38
rs772651740 Chr.8: 65704908-65704908 on Build GRCh38
rs772650797 Chr.8: 65704888-65704888 on Build GRCh38
rs770953543 Chr.8: 65704900-65704900 on Build GRCh38
rs909142191 Chr.8: 65704882-65704882 on Build GRCh38
rs139624623 Chr.8: 65704814-65704814 on Build GRCh38
rs775848639 Chr.8: 65704894-65704894 on Build GRCh38
rs1036963841 Chr.8: 65704956-65704956 on Build GRCh38
rs781077499 Chr.8: 65704848-65704848 on Build GRCh38
rs768872348 Chr.8: 65704900-65704900 on Build GRCh38
rs979111734 Chr.8: 65704866-65704866 on Build GRCh38
rs887859188 Chr.8: 65705009-65705009 on Build GRCh38
rs747620280 Chr.8: 65704898-65704898 on Build GRCh38
rs897803333 Chr.8: 65704987-65704987 on Build GRCh38
rs371050522 Chr.8: 65704909-65704909 on Build GRCh38
rs779099964 Chr.8: 65704854-65704854 on Build GRCh38

View Full Product Details