Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 3320236-3320475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MXRA5
Entrez Gene ID
25878
Gene Name
matrix remodeling associated 5
Gene Aliases
-
UniGene
Hs.369422
Gene Chromosome Location
Chr.X: 3308565-3347098 on Build GRCh38
Transcripts
NM_015419.3 XM_005274485.2

SNP Detail

SNP ID Chromosome Location
rs757052581 Chr.X: 3320414-3320414 on Build GRCh38
rs745852995 Chr.X: 3320283-3320283 on Build GRCh38
rs773306775 Chr.X: 3320327-3320327 on Build GRCh38
rs1011174456 Chr.X: 3320359-3320359 on Build GRCh38
rs868250427 Chr.X: 3320437-3320437 on Build GRCh38
rs982684282 Chr.X: 3320382-3320382 on Build GRCh38
rs779975747 Chr.X: 3320446-3320446 on Build GRCh38
rs79552860 Chr.X: 3320435-3320435 on Build GRCh38
rs890535898 Chr.X: 3320265-3320265 on Build GRCh38
rs760117677 Chr.X: 3320391-3320391 on Build GRCh38
rs376762981 Chr.X: 3320298-3320298 on Build GRCh38
rs763762016 Chr.X: 3320408-3320408 on Build GRCh38
rs747125005 Chr.X: 3320427-3320427 on Build GRCh38
rs142902268 Chr.X: 3320330-3320330 on Build GRCh38
rs760162200 Chr.X: 3320349-3320349 on Build GRCh38
rs762235263 Chr.X: 3320374-3320374 on Build GRCh38
rs34070401 Chr.X: 3320389-3320390 on Build GRCh38
rs866581403 Chr.X: 3320302-3320302 on Build GRCh38
rs778773109 Chr.X: 3320281-3320281 on Build GRCh38
rs774980117 Chr.X: 3320340-3320340 on Build GRCh38
rs112302509 Chr.X: 3320417-3320417 on Build GRCh38
rs753454467 Chr.X: 3320409-3320409 on Build GRCh38
rs767759204 Chr.X: 3320437-3320445 on Build GRCh38
rs758385816 Chr.X: 3320432-3320432 on Build GRCh38
rs966926453 Chr.X: 3320376-3320376 on Build GRCh38
rs748301955 Chr.X: 3320306-3320306 on Build GRCh38
rs777185808 Chr.X: 3320368-3320368 on Build GRCh38
rs867779664 Chr.X: 3320295-3320295 on Build GRCh38
rs957998828 Chr.X: 3320331-3320331 on Build GRCh38
rs200611623 Chr.X: 3320332-3320332 on Build GRCh38
rs145010278 Chr.X: 3320297-3320297 on Build GRCh38
rs369769079 Chr.X: 3320313-3320313 on Build GRCh38
rs757033785 Chr.X: 3320273-3320273 on Build GRCh38
rs866073929 Chr.X: 3320418-3320418 on Build GRCh38

View Full Product Details