Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 32017132-32017391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF5B
Entrez Gene ID
3799
Gene Name
kinesin family member 5B
Gene Aliases
HEL-S-61, KINH, KNS, KNS1, UKHC
UniGene
Hs.327736
Gene Chromosome Location
Chr.10: 32009010-32056443 on Build GRCh38
Transcripts
NM_004521.2 XM_017016224.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
684480 c.2727G>A p.K909K Chr.10: 32017177-32017177 on Build GRCh38
5428757 c.2685T>A p.Y895* Chr.10: 32017219-32017219 on Build GRCh38
917752 c.2656G>T p.E886* Chr.10: 32017248-32017248 on Build GRCh38
917751 c.2702G>A p.R901H Chr.10: 32017202-32017202 on Build GRCh38
7112857 c.2667delT p.R890fs*10 Chr.10: 32017237-32017237 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1021461248 Chr.10: 32017165-32017165 on Build GRCh38
rs903863790 Chr.10: 32017199-32017199 on Build GRCh38
rs148741299 Chr.10: 32017181-32017181 on Build GRCh38
rs747074223 Chr.10: 32017180-32017180 on Build GRCh38
rs751428790 Chr.10: 32017279-32017279 on Build GRCh38
rs1008333486 Chr.10: 32017186-32017186 on Build GRCh38
rs773585260 Chr.10: 32017230-32017230 on Build GRCh38
rs141491660 Chr.10: 32017224-32017224 on Build GRCh38
rs370648605 Chr.10: 32017365-32017365 on Build GRCh38
rs11539712 Chr.10: 32017167-32017167 on Build GRCh38
rs764985083 Chr.10: 32017254-32017254 on Build GRCh38
rs768225070 Chr.10: 32017237-32017237 on Build GRCh38
rs750467260 Chr.10: 32017191-32017192 on Build GRCh38
rs150818260 Chr.10: 32017308-32017308 on Build GRCh38
rs749660381 Chr.10: 32017326-32017326 on Build GRCh38
rs749798990 Chr.10: 32017231-32017231 on Build GRCh38
rs377196810 Chr.10: 32017293-32017293 on Build GRCh38
rs779577244 Chr.10: 32017334-32017334 on Build GRCh38
rs547388371 Chr.10: 32017235-32017235 on Build GRCh38
rs761519908 Chr.10: 32017249-32017249 on Build GRCh38
rs200746362 Chr.10: 32017352-32017352 on Build GRCh38
rs780042723 Chr.10: 32017177-32017177 on Build GRCh38
rs766332268 Chr.10: 32017278-32017278 on Build GRCh38
rs1027532542 Chr.10: 32017348-32017348 on Build GRCh38
rs750214006 Chr.10: 32017266-32017266 on Build GRCh38
rs758329459 Chr.10: 32017163-32017163 on Build GRCh38
rs755060428 Chr.10: 32017282-32017282 on Build GRCh38
rs768127731 Chr.10: 32017181-32017181 on Build GRCh38
rs992235645 Chr.10: 32017227-32017227 on Build GRCh38
rs763280306 Chr.10: 32017261-32017261 on Build GRCh38
rs776449498 Chr.10: 32017238-32017238 on Build GRCh38
rs756328599 Chr.10: 32017317-32017317 on Build GRCh38
rs755716349 Chr.10: 32017203-32017203 on Build GRCh38
rs964545969 Chr.10: 32017247-32017247 on Build GRCh38
rs760268366 Chr.10: 32017236-32017236 on Build GRCh38
rs746329885 Chr.10: 32017341-32017341 on Build GRCh38
rs762852206 Chr.10: 32017270-32017270 on Build GRCh38
rs565996129 Chr.10: 32017321-32017321 on Build GRCh38
rs771134833 Chr.10: 32017330-32017330 on Build GRCh38
rs1031402203 Chr.10: 32017223-32017223 on Build GRCh38

View Full Product Details