Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 38163617-38163888 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LSM1
Entrez Gene ID
27257
Gene Name
LSM1 homolog, mRNA degradation associated
Gene Aliases
CASM, YJL124C
UniGene
Hs.425311
Gene Chromosome Location
Chr.8: 38163321-38176730 on Build GRCh38
Transcripts
NM_014462.2 NR_045492.1 NR_045493.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2960141 c.380C>T p.A127V Chr.8: 38163692-38163692 on Build GRCh38
1099551 c.263A>G p.Q88R Chr.8: 38163809-38163809 on Build GRCh38
4570859 c.334T>C p.L112L Chr.8: 38163738-38163738 on Build GRCh38
3648809 c.394G>C p.E132Q Chr.8: 38163678-38163678 on Build GRCh38
7319331 c.308A>C p.Q103P Chr.8: 38163764-38163764 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758491210 Chr.8: 38163726-38163726 on Build GRCh38
rs745440412 Chr.8: 38163674-38163676 on Build GRCh38
rs569027177 Chr.8: 38163807-38163807 on Build GRCh38
rs1045345664 Chr.8: 38163739-38163739 on Build GRCh38
rs769434967 Chr.8: 38163683-38163687 on Build GRCh38
rs776349391 Chr.8: 38163837-38163837 on Build GRCh38
rs370074152 Chr.8: 38163653-38163653 on Build GRCh38
rs539421938 Chr.8: 38163852-38163852 on Build GRCh38
rs754371640 Chr.8: 38163670-38163670 on Build GRCh38
rs11551204 Chr.8: 38163773-38163773 on Build GRCh38
rs773956422 Chr.8: 38163784-38163784 on Build GRCh38
rs372428495 Chr.8: 38163772-38163772 on Build GRCh38
rs757496594 Chr.8: 38163648-38163648 on Build GRCh38
rs760456370 Chr.8: 38163813-38163813 on Build GRCh38
rs750856441 Chr.8: 38163725-38163725 on Build GRCh38
rs780355621 Chr.8: 38163735-38163735 on Build GRCh38
rs752184193 Chr.8: 38163651-38163651 on Build GRCh38
rs907657329 Chr.8: 38163649-38163649 on Build GRCh38
rs936889222 Chr.8: 38163781-38163781 on Build GRCh38
rs779139978 Chr.8: 38163715-38163715 on Build GRCh38
rs745676237 Chr.8: 38163801-38163801 on Build GRCh38
rs757876995 Chr.8: 38163703-38163703 on Build GRCh38
rs917561702 Chr.8: 38163713-38163713 on Build GRCh38
rs780980149 Chr.8: 38163636-38163637 on Build GRCh38
rs758812966 Chr.8: 38163857-38163857 on Build GRCh38
rs750687126 Chr.8: 38163855-38163855 on Build GRCh38
rs747041639 Chr.8: 38163744-38163744 on Build GRCh38
rs763669019 Chr.8: 38163820-38163820 on Build GRCh38
rs961755643 Chr.8: 38163696-38163696 on Build GRCh38
rs764644449 Chr.8: 38163654-38163654 on Build GRCh38
rs781288230 Chr.8: 38163763-38163763 on Build GRCh38
rs755027535 Chr.8: 38163755-38163755 on Build GRCh38
rs775307737 Chr.8: 38163810-38163810 on Build GRCh38
rs765787346 Chr.8: 38163854-38163854 on Build GRCh38

View Full Product Details