Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 98133454-98133705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR5H1
Entrez Gene ID
26341
Gene Name
olfactory receptor family 5 subfamily H member 1
Gene Aliases
HSHTPCRX14, HTPCRX14
UniGene
Hs.537383
Gene Chromosome Location
Chr.3: 98132698-98133639 on Build GRCh38
Transcripts
NM_001005338.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2950066 c.895G>A p.V299I Chr.3: 98133592-98133592 on Build GRCh38
5966085 c.891C>G p.V297V Chr.3: 98133588-98133588 on Build GRCh38
731339 c.935C>G p.S312* Chr.3: 98133632-98133632 on Build GRCh38
5950308 c.923A>T p.H308L Chr.3: 98133620-98133620 on Build GRCh38
7276416 c.818T>C p.V273A Chr.3: 98133515-98133515 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542298979 Chr.3: 98133533-98133533 on Build GRCh38
rs549706159 Chr.3: 98133646-98133646 on Build GRCh38
rs576551305 Chr.3: 98133513-98133513 on Build GRCh38
rs533241539 Chr.3: 98133640-98133640 on Build GRCh38
rs527463332 Chr.3: 98133610-98133610 on Build GRCh38
rs562175052 Chr.3: 98133557-98133557 on Build GRCh38
rs763936444 Chr.3: 98133623-98133623 on Build GRCh38
rs200536795 Chr.3: 98133648-98133648 on Build GRCh38
rs530358694 Chr.3: 98133630-98133630 on Build GRCh38
rs772430800 Chr.3: 98133589-98133589 on Build GRCh38
rs779355523 Chr.3: 98133661-98133661 on Build GRCh38
rs566338612 Chr.3: 98133480-98133480 on Build GRCh38
rs756745869 Chr.3: 98133528-98133528 on Build GRCh38
rs745373835 Chr.3: 98133537-98133537 on Build GRCh38
rs761949326 Chr.3: 98133496-98133496 on Build GRCh38
rs770967072 Chr.3: 98133677-98133677 on Build GRCh38
rs541245162 Chr.3: 98133622-98133622 on Build GRCh38
rs759454757 Chr.3: 98133508-98133508 on Build GRCh38
rs750155763 Chr.3: 98133612-98133612 on Build GRCh38
rs140293806 Chr.3: 98133485-98133485 on Build GRCh38
rs769126214 Chr.3: 98133657-98133657 on Build GRCh38
rs755606881 Chr.3: 98133642-98133642 on Build GRCh38
rs765181736 Chr.3: 98133612-98133612 on Build GRCh38
rs770278660 Chr.3: 98133544-98133544 on Build GRCh38
rs150453860 Chr.3: 98133535-98133535 on Build GRCh38
rs1004230976 Chr.3: 98133572-98133573 on Build GRCh38
rs777374780 Chr.3: 98133520-98133520 on Build GRCh38
rs867848789 Chr.3: 98133669-98133669 on Build GRCh38
rs1033593914 Chr.3: 98133634-98133634 on Build GRCh38
rs772374647 Chr.3: 98133667-98133667 on Build GRCh38
rs770047951 Chr.3: 98133484-98133484 on Build GRCh38
rs767752384 Chr.3: 98133501-98133501 on Build GRCh38
rs748354794 Chr.3: 98133645-98133645 on Build GRCh38
rs769179448 Chr.3: 98133563-98133563 on Build GRCh38
rs190397100 Chr.3: 98133491-98133491 on Build GRCh38
rs138973744 Chr.3: 98133492-98133492 on Build GRCh38
rs762196178 Chr.3: 98133573-98133573 on Build GRCh38
rs564155002 Chr.3: 98133635-98133635 on Build GRCh38
rs376802329 Chr.3: 98133591-98133591 on Build GRCh38
rs778078239 Chr.3: 98133641-98133641 on Build GRCh38
rs760777784 Chr.3: 98133676-98133676 on Build GRCh38
rs748539535 Chr.3: 98133662-98133662 on Build GRCh38
rs751264873 Chr.3: 98133525-98133525 on Build GRCh38
rs751206050 Chr.3: 98133628-98133628 on Build GRCh38
rs970766809 Chr.3: 98133627-98133627 on Build GRCh38
rs774689923 Chr.3: 98133567-98133567 on Build GRCh38
rs781469082 Chr.3: 98133619-98133619 on Build GRCh38
rs758159596 Chr.3: 98133514-98133514 on Build GRCh38
rs267599953 Chr.3: 98133556-98133556 on Build GRCh38
rs111392904 Chr.3: 98133503-98133503 on Build GRCh38
rs758145105 Chr.3: 98133611-98133612 on Build GRCh38
rs780443361 Chr.3: 98133553-98133553 on Build GRCh38
rs778856905 Chr.3: 98133550-98133550 on Build GRCh38

View Full Product Details