Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 42764082-42764307 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OXER1
Entrez Gene ID
165140
Gene Name
oxoeicosanoid receptor 1
Gene Aliases
GPCR, GPR170, TG1019
UniGene
Hs.168439
Gene Chromosome Location
Chr.2: 42762499-42764261 on Build GRCh38
Transcripts
NM_148962.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6217464 c.19G>A p.G7S Chr.2: 42764161-42764161 on Build GRCh38
1293988 c.13C>T p.R5C Chr.2: 42764167-42764167 on Build GRCh38
5719588 c.87C>T p.N29N Chr.2: 42764093-42764093 on Build GRCh38
6010096 c.93C>G p.L31L Chr.2: 42764087-42764087 on Build GRCh38
6542538 c.71G>A p.G24D Chr.2: 42764109-42764109 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368634132 Chr.2: 42764191-42764191 on Build GRCh38
rs559631497 Chr.2: 42764201-42764201 on Build GRCh38
rs1017378662 Chr.2: 42764118-42764118 on Build GRCh38
rs773355020 Chr.2: 42764144-42764144 on Build GRCh38
rs986251099 Chr.2: 42764279-42764279 on Build GRCh38
rs539369957 Chr.2: 42764266-42764266 on Build GRCh38
rs201353687 Chr.2: 42764214-42764214 on Build GRCh38
rs760846891 Chr.2: 42764149-42764149 on Build GRCh38
rs755609616 Chr.2: 42764180-42764180 on Build GRCh38
rs765531235 Chr.2: 42764221-42764221 on Build GRCh38
rs976498036 Chr.2: 42764272-42764272 on Build GRCh38
rs747961065 Chr.2: 42764204-42764204 on Build GRCh38
rs770883225 Chr.2: 42764210-42764210 on Build GRCh38
rs914649054 Chr.2: 42764282-42764282 on Build GRCh38
rs371146710 Chr.2: 42764128-42764128 on Build GRCh38
rs953502745 Chr.2: 42764278-42764278 on Build GRCh38
rs1056102251 Chr.2: 42764192-42764192 on Build GRCh38
rs973236844 Chr.2: 42764138-42764138 on Build GRCh38
rs766621052 Chr.2: 42764151-42764151 on Build GRCh38
rs775020395 Chr.2: 42764109-42764109 on Build GRCh38
rs548962568 Chr.2: 42764122-42764122 on Build GRCh38
rs753131710 Chr.2: 42764170-42764170 on Build GRCh38
rs749030249 Chr.2: 42764114-42764114 on Build GRCh38
rs755270340 Chr.2: 42764158-42764158 on Build GRCh38
rs749879038 Chr.2: 42764178-42764178 on Build GRCh38
rs759631327 Chr.2: 42764215-42764215 on Build GRCh38
rs748868056 Chr.2: 42764198-42764198 on Build GRCh38
rs774249867 Chr.2: 42764115-42764115 on Build GRCh38
rs371144324 Chr.2: 42764203-42764203 on Build GRCh38
rs923257904 Chr.2: 42764277-42764277 on Build GRCh38
rs938695342 Chr.2: 42764189-42764189 on Build GRCh38
rs778258014 Chr.2: 42764176-42764176 on Build GRCh38
rs140987475 Chr.2: 42764166-42764166 on Build GRCh38
rs1014813044 Chr.2: 42764231-42764231 on Build GRCh38
rs374249711 Chr.2: 42764208-42764208 on Build GRCh38
rs772073842 Chr.2: 42764205-42764205 on Build GRCh38
rs929151435 Chr.2: 42764162-42764162 on Build GRCh38
rs780809061 Chr.2: 42764147-42764147 on Build GRCh38
rs753999081 Chr.2: 42764155-42764155 on Build GRCh38
rs374208009 Chr.2: 42764174-42764174 on Build GRCh38
rs192426679 Chr.2: 42764283-42764283 on Build GRCh38
rs371620646 Chr.2: 42764225-42764225 on Build GRCh38
rs547304533 Chr.2: 42764223-42764223 on Build GRCh38
rs570599775 Chr.2: 42764262-42764262 on Build GRCh38
rs760758899 Chr.2: 42764209-42764209 on Build GRCh38

View Full Product Details